Assay Details
Target Gene Details
Entrez Gene ID: | 6651 |
Gene Name: | SON DNA binding protein |
Gene Aliases: |
BASS1, C21orf50, DBP-5, NREBP, SON3 |
Location: |
Chr.21:33543038-33577514 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SON | NM_001291411.1 | 3 | 2067 | NP_001278340.1 |
NM_001291412.1 | NP_001278341.1 | |||
NM_032195.2 | 3 | 2067 | NP_115571.2 | |
NM_138927.2 | 3 | 2067 | NP_620305.2 | |
NR_103797.1 | 3 | 2067 | ||
AB028942.2 | 3 | 2036 | ||
AF380179.1 | AAL34497.1 | |||
AF380180.1 | 3 | 2041 | AAL34498.1 | |
AF380181.1 | 3 | 2041 | AAL34499.1 | |
AF380183.1 | 3 | 2041 | AAL34501.1 | |
AF380184.1 | 3 | 2041 | AAL34502.1 | |
AF435977.1 | AAL30810.1 | |||
AK074269.1 | 1 | 1028 | ||
AK091233.1 | ||||
AK127947.1 | BAC87202.1 | |||
AK128071.1 | 1 | 1014 | ||
AK307612.1 | 3 | 2067 | ||
AY026895.1 | 3 | 2061 | AAK07692.1 | |
BC002422.2 | 1 | 531 | AAH02422.1 | |
X63753.1 | CAA45282.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1115615 | Chr.21:33548994 - 33557394 on Build GRCh38 | Duplication | MIR6501 SON |
nsv1078502 | Chr.21:33549893 - 33553894 on Build GRCh38 | Duplication | MIR6501 SON |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |