Assay Details
Target Gene Details
Entrez Gene ID: | 29947 |
Gene Name: | DNA methyltransferase 3 like |
Gene Aliases: |
- |
Location: |
Chr.21:44246339-44262216 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DNMT3L | NM_013369.3 | NP_037501.2 | ||
NM_175867.2 | NP_787063.1 | |||
AF194032.1 | AAF05812.1 | |||
BC002560.2 | AAH02560.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv587741 | Chr.21:44248288 - 44263467 on Build GRCh38 | Loss |
![]() ![]() |
nsv953645 | Chr.21:44156018 - 44264017 on Build GRCh38 | Deletion |
![]() ![]() ![]() |
nsv587743 | Chr.21:44255994 - 44263467 on Build GRCh38 | Gain |
![]() |
esv2723638 | Chr.21:44249174 - 44593709 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv587740 | Chr.21:44209682 - 44429981 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
esv33817 | Chr.21:43749531 - 44647625 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)