Assay Details
Target Gene Details
Entrez Gene ID: | 4335 |
Gene Name: | MAX network transcriptional repressor |
Gene Aliases: |
MAD6, MXD6, ROX, bHLHd3 |
Location: |
Chr.17:2384060-2400964 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MNT | NM_020310.2 | 6 | 3399 | NP_064706.1 |
XM_011523868.2 | 6 | 3336 | XP_011522170.1 | |
XM_011523869.1 | 3 | 2433 | XP_011522171.1 | |
XM_017024654.1 | 3 | 5045 | XP_016880143.1 | |
BC117563.1 | 6 | 3149 | AAI17564.1 | |
BM820333.1 | ||||
BQ675978.1 | 1 | 628 | ||
X96401.1 | 6 | 3359 | CAA65265.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv833338 | Chr.17:2363607 - 2592205 on Build GRCh38 | Loss | LOC101927864 MNT SGSM2 LOC284009 METTL16 |
nsv470569 | Chr.17:2359409 - 2395680 on Build GRCh38 | Loss | LOC101927864 MNT SGSM2 |
nsv1063109 | Chr.17:2342302 - 2591288 on Build GRCh38 | Gain | LOC101927864 MNT SGSM2 LOC284009 METTL16 |
nsv469765 | Chr.17:2213155 - 2400446 on Build GRCh38 | Loss | SMG6 LOC101927864 MNT SNORD91A LOC101927839 SGSM2 SNORD91B TSR1 SRR |
nsv952104 | Chr.17:2337907 - 2399506 on Build GRCh38 | Deletion | LOC101927864 MNT SGSM2 |
nsv1160444 | Chr.17:2377640 - 2402666 on Build GRCh38 | Deletion | MNT SGSM2 |
nsv1160443 | Chr.17:2346610 - 2406836 on Build GRCh38 | Deletion | LOC101927864 MNT SGSM2 |
nsv1066475 | Chr.17:2359823 - 2390866 on Build GRCh38 | Loss | LOC101927864 MNT SGSM2 |
dgv3083n100 | Chr.17:2351844 - 2409068 on Build GRCh38 | Gain | LOC101927864 MNT SGSM2 LOC284009 |
nsv833336 | Chr.17:2176922 - 2400456 on Build GRCh38 | Loss | SMG6 LOC101927864 MNT SNORD91A LOC101927839 SGSM2 SNORD91B TSR1 SRR |
nsv574224 | Chr.17:2269769 - 2423186 on Build GRCh38 | Gain | SMG6 LOC101927864 MNT SNORD91A SGSM2 LOC284009 SNORD91B TSR1 SRR METTL16 |
nsv517489 | Chr.17:2344688 - 2395680 on Build GRCh38 | Loss | LOC101927864 MNT SGSM2 |
nsv1055225 | Chr.17:2343962 - 2389565 on Build GRCh38 | Loss | LOC101927864 MNT SGSM2 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |