Assay Details
Target Gene Details
Entrez Gene ID: | 23493 |
Gene Name: | hes related family bHLH transcription factor with YRPW motif 2 |
Gene Aliases: |
CHF1, GRIDLOCK, GRL, HERP1, HESR2, HRT2, bHLHb32 |
Location: |
Chr.6:125747592-125762243 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
HEY2 | NM_012259.2 | 5 | 858 | NP_036391.1 |
XM_017010627.1 | 5 | 2242 | XP_016866116.1 | |
XM_017010628.1 | 6 | 1940 | XP_016866117.1 | |
XM_017010629.1 | 5 | 719 | XP_016866118.1 | |
AB044755.1 | 5 | 763 | BAA96781.1 | |
AF173901.1 | 5 | 826 | AAF20173.1 | |
AF232238.1 | 5 | 777 | AAF37296.1 | |
AF237949.1 | 5 | 734 | AAF44781.1 | |
AF311884.1 | 5 | 667 | AAG31157.1 | |
AJ249545.1 | 5 | 717 | CAB56839.1 | |
AK289943.1 | 5 | 776 | ||
AK303890.1 | 5 | 680 | ||
BC007707.2 | 5 | 756 | AAH07707.1 | |
BT020067.1 | 5 | 667 | AAV38870.1 | |
BT020068.1 | 5 | 667 | AAV38871.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |