Assay Details
Target Gene Details
Entrez Gene ID: | 51380 |
Gene Name: | cysteine sulfinic acid decarboxylase |
Gene Aliases: |
CSD, PCAP |
Location: |
Chr.12:53157663-53180909 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 14 - Exon 14 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv832417 | Chr.12:53075874 - 53254231 on Build GRCh38 | Gain | ZNF740 ITGB7 CSAD MFSD5 SPRYD3 SOAT2 IGFBP6 RARG |
nsv558994 | Chr.12:53103618 - 53277765 on Build GRCh38 | Loss | ESPL1 ZNF740 ITGB7 CSAD MFSD5 SOAT2 RARG |
dgv2645n54 | Chr.12:53099603 - 53227927 on Build GRCh38 | Loss | ZNF740 ITGB7 CSAD SOAT2 IGFBP6 RARG |
nsv517066 | Chr.12:53006394 - 53340722 on Build GRCh38 | Loss | LOC283335 SP7 CSAD C12orf10 SPRYD3 AAAS TNS2 SOAT2 IGFBP6 RARG MIR6757 ESPL1 ZNF740 ITGB7 PFDN5 EIF4B MFSD5 |
nsv469404 | Chr.12:53124036 - 53227927 on Build GRCh38 | Loss | ZNF740 ITGB7 CSAD SOAT2 RARG |
dgv2644n54 | Chr.12:53064109 - 53211560 on Build GRCh38 | Loss | ZNF740 ITGB7 CSAD SPRYD3 TNS2 SOAT2 IGFBP6 RARG |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |