Assay Details
Target Gene Details
Entrez Gene ID: | 51657 |
Gene Name: | serine/threonine/tyrosine interacting-like 1 |
Gene Aliases: |
DUSP24, MK-STYX, MKSTYX |
Location: |
Chr.7:75995820-76048709 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv428173 | Chr.7:75934531 - 76173362 on Build GRCh38 | Gain | POR GTF2IP7 LOC107986711 STYXL1 TMEM120A SNORA14A MDH2 |
nsv528969 | Chr.7:75074053 - 76920728 on Build GRCh38 | Gain | RHBDD2 POR ZP3 LOC541473 STAG3L1 PMS2P2 LOC100133091 PMS2P3 NCF1C PMS2P10 DTX2 HSPB1 LOC107986711 CCL26 SPDYE5 POM121C SNORA14A MDH2 HIP1 SPDYE16 CCL24 NSUN5P1 POMZP3 SSC4D LOC441259 LOC105375355 GTF2IP7 FDPSP2 GTF2IRD2B STYXL1 TMEM120A SRRM3 GTF2IP1 TRIM73 YWHAG MIR4651 |
esv3613742 | Chr.7:75912784 - 76036530 on Build GRCh38 | Loss | POR STYXL1 TMEM120A SNORA14A MIR4651 |
esv3613749 | Chr.7:76009631 - 76035709 on Build GRCh38 | Loss | STYXL1 |
dgv120n16 | Chr.7:75440769 - 76641919 on Build GRCh38 | Deletion | RHBDD2 POR ZP3 LOC100133091 PMS2P3 DTX2 HSPB1 LOC107986711 CCL26 SPDYE5 POM121C SNORA14A MDH2 HIP1 SPDYE16 CCL24 POMZP3 SSC4D LOC105375355 GTF2IP7 FDPSP2 STYXL1 TMEM120A SRRM3 YWHAG MIR4651 |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |