Assay Details
Target Gene Details
Entrez Gene ID: | 51706 |
Gene Name: | cytochrome b5 reductase 1 |
Gene Aliases: |
B5R.1, B5R1, B5R2, NQO3A2, humb5R2 |
Location: |
Chr.1:202961873-202967276 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CYB5R1 | NM_016243.2 | NP_057327.2 | ||
AF087912.1 | AAP97209.1 | |||
AF093822.1 | AAP97218.1 | |||
AF125533.1 | AAF17227.1 | |||
AF169481.1 | AAF06147.1 | |||
AK074654.1 | BAC11115.1 | |||
AK075304.1 | ||||
AK223416.1 | BAD97136.1 | |||
AK295902.1 | ||||
AK313333.1 | ||||
AY359026.1 | AAQ89385.1 | |||
BC018732.1 | AAH18732.1 | |||
BC127945.1 |
More Information
Set Membership: |
Intragenic Non-exonic non-DGV Variation |