Assay Details
Target Gene Details
Entrez Gene ID: | 8402 |
Gene Name: | solute carrier family 25 member 11 |
Gene Aliases: |
OGC, SLC20A4 |
Location: |
Chr.17:4937130-4940167 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SLC25A11 | NM_001165417.1 | NP_001158889.1 | ||
NM_001165418.1 | NP_001158890.1 | |||
NM_003562.4 | NP_003553.2 | |||
AF070548.1 | AAC28637.1 | |||
AK290376.1 | ||||
AK313010.1 | ||||
BC006508.2 | AAH06508.1 | |||
BC006519.2 | AAH06519.1 | |||
BC016294.2 | AAH16294.1 | |||
BC017170.2 | AAH17170.1 | |||
BI752999.1 | ||||
BM047391.1 | ||||
CB155476.1 | ||||
CR456834.1 | CAG33115.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv833350 | Chr.17:4884713 - 5060776 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
esv3572365 | Chr.17:4874864 - 4942141 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() |
nsv833349 | Chr.17:4853631 - 4991300 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv574275 | Chr.17:4875192 - 4946479 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv833347 | Chr.17:4785297 - 4955981 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
dgv3088n100 | Chr.17:4868317 - 4942562 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() |
nsv574273 | Chr.17:4585589 - 5460332 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)