Assay Details
Target Gene Details
Entrez Gene ID: | 79654 |
Gene Name: | HECT domain E3 ubiquitin protein ligase 3 |
Gene Aliases: |
- |
Location: |
Chr.1:45002548-45011355 on Build GRCh38 |
Assay Gene Location: | Within Exon 21 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
HECTD3 | NM_024602.5 | 21 | 2751 | NP_078878.3 |
XM_011542140.2 | 19 | 2148 | XP_011540442.1 | |
AK056545.1 | 19 | 2102 | ||
AK056873.1 | 13 | 1645 | ||
AK091583.1 | 6 | 1500 | ||
AK096462.1 | 21 | 2630 | ||
AK301124.1 | ||||
BC019105.2 | 6 | 710 | ||
BC126352.1 | 18 | 2028 | ||
BC130470.1 | 18 | 2028 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv950706 | Chr.1:45001329 - 45011128 on Build GRCh38 | Deletion | HECTD3 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |