Assay Details
Target Gene Details
Entrez Gene ID: | 8711 |
Gene Name: | tyrosine kinase non receptor 1 |
Gene Aliases: |
- |
Location: |
Chr.17:7380559-7389774 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TNK1 | NM_001251902.1 | NP_001238831.1 | ||
NM_003985.4 | NP_003976.2 | |||
XM_011524045.2 | XP_011522347.1 | |||
XM_011524046.2 | XP_011522348.1 | |||
XM_011524047.2 | XP_011522349.1 | |||
XM_011524048.1 | XP_011522350.1 | |||
AK225403.1 | ||||
AK310684.1 | ||||
BC035782.1 | AAH35782.1 | |||
U43408.1 | AAC50427.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2422288 | Chr.17:6789933 - 7646709 on Build GRCh38 | Deletion | TNFSF12-TNFSF13 MIR324 SAT2 TEKT1 MIR497 ACADVL CD68 ALOX12 SNORA67 CLEC10A TNFSF12 DVL2 KCTD11 RNASEK TMEM102 CLDN7 SLC35G6 SNORD10 SOX15 C17orf49 BCL6B FGF11 ASGR2 PLSCR3 ELP5 LOC107983988 SNORA48 C17orf74 FXR2 EIF5A ATP1B2 MPDU1 ALOX12P2 SLC16A13 ASGR1 MIR497HG PHF23 NLGN2 SHBG TMEM256-PLSCR3 CTDNEP1 SPEM1 LOC100996842 TNK1 SLC2A4 GABARAP GPS2 RNASEK-C17orf49 ACAP1 SENP3 SLC16A11 POLR2A ZBTB4 ALOX12-AS1 TMEM95 MIR195 EIF4A1 NEURL4 FBXO39 CHRNB1 YBX2 SENP3-EIF4A1 TMEM256 DLG4 TNFSF13 |
nsv523672 | Chr.17:7379825 - 7549660 on Build GRCh38 | Loss | TNFSF12-TNFSF13 POLR2A ZBTB4 NLGN2 TMEM256-PLSCR3 SPEM1 TNK1 TNFSF12 FGF11 PLSCR3 TMEM102 LOC107983988 CHRNB1 C17orf74 SLC35G6 TMEM256 |
nsv952118 | Chr.17:7174582 - 7456981 on Build GRCh38 | Deletion | MIR324 PHF23 NLGN2 TMEM256-PLSCR3 CTDNEP1 ACADVL SPEM1 TNK1 SLC2A4 DVL2 KCTD11 GABARAP TMEM102 GPS2 CLDN7 ACAP1 TMEM95 FGF11 NEURL4 PLSCR3 ELP5 LOC107983988 CHRNB1 C17orf74 YBX2 EIF5A TMEM256 DLG4 ASGR1 |
nsv574322 | Chr.17:7010333 - 7689462 on Build GRCh38 | Loss | TNFSF12-TNFSF13 MIR324 SAT2 MIR497 ACADVL CD68 ALOX12 SNORA67 CLEC10A TNFSF12 DVL2 KCTD11 RNASEK TMEM102 CLDN7 SLC35G6 SNORD10 SOX15 WRAP53 C17orf49 BCL6B FGF11 ASGR2 PLSCR3 ELP5 LOC107983988 SNORA48 C17orf74 FXR2 EIF5A ATP1B2 MPDU1 SLC16A13 ASGR1 MIR497HG PHF23 NLGN2 SHBG TMEM256-PLSCR3 CTDNEP1 SPEM1 LOC100996842 TNK1 SLC2A4 GABARAP GPS2 RNASEK-C17orf49 ACAP1 TP53 SENP3 SLC16A11 POLR2A ZBTB4 ALOX12-AS1 TMEM95 MIR195 EIF4A1 NEURL4 CHRNB1 YBX2 SENP3-EIF4A1 TMEM256 DLG4 TNFSF13 |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |