Assay Details
Target Gene Details
Entrez Gene ID: | 51455 |
Gene Name: | REV1, DNA directed polymerase |
Gene Aliases: |
AIBP80, REV1L |
Location: |
Chr.2:99400475-99490123 on Build GRCh38 |
Assay Gene Location: | Within Exon 28 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
REV1 | NM_001037872.2 | 23 | 4013 | NP_001032961.1 |
NM_001321454.1 | 24 | 4124 | NP_001308383.1 | |
NM_001321455.1 | 24 | 4170 | NP_001308384.1 | |
NM_001321458.1 | 24 | 4140 | NP_001308387.1 | |
NM_001321459.1 | 24 | 4216 | NP_001308388.1 | |
NM_001321460.1 | 24 | 4213 | NP_001308389.1 | |
NM_016316.3 | 23 | 4016 | NP_057400.1 | |
NR_135649.1 | 23 | 4510 | ||
NR_135650.1 | 23 | 4467 | ||
NR_135651.1 | 22 | 3847 | ||
NR_135652.1 | 22 | 4313 | ||
NR_135653.1 | 22 | 3850 | ||
XM_011511341.2 | 17 | 2633 | XP_011509643.1 | |
XM_017004309.1 | 24 | 4537 | XP_016859798.1 | |
XM_017004310.1 | 24 | 3903 | XP_016859799.1 | |
XM_017004311.1 | 24 | 3903 | XP_016859800.1 | |
XM_017004312.1 | 24 | 4116 | XP_016859801.1 | |
XM_017004313.1 | 23 | 5742 | XP_016859802.1 | |
XM_017004314.1 | 20 | 3393 | XP_016859803.1 | |
XM_017004315.1 | 20 | 3377 | XP_016859804.1 | |
AB047646.1 | 23 | 3938 | BAB21441.1 | |
AF151538.1 | 23 | 3969 | AAF06731.1 | |
AF206019.1 | 23 | 3978 | AAF18986.1 | |
AF357886.1 | 23 | 3934 | AAK43708.1 | |
AJ131720.1 | 15 | 2335 | CAB38231.1 | |
BC037734.1 | 23 | 4448 | AAH37734.1 | |
BC130411.1 | ||||
CA951106.1 | 2 | 520 | ||
CK300059.1 | 1 | 299 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv4032n100 | Chr.2:99364388 - 99481422 on Build GRCh38 | Gain | REV1 EIF5B |
nsv524466 | Chr.2:99335623 - 99415089 on Build GRCh38 | Loss | TXNDC9 REV1 EIF5B |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |