Assay Details
Target Gene Details
Entrez Gene ID: | 8372 |
Gene Name: | hyaluronoglucosaminidase 3 |
Gene Aliases: |
HYAL-3, LUCA-3, LUCA3 |
Location: |
Chr.3:50292828-50299468 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
HYAL3 | NM_001200029.1 | 4 | 1545 | NP_001186958.1 |
NM_001200030.1 | 3 | 1669 | NP_001186959.1 | |
NM_001200031.1 | 4 | 993 | NP_001186960.1 | |
NM_001200032.1 | 3 | 903 | NP_001186961.1 | |
NM_003549.3 | 4 | 1759 | NP_003540.2 | |
AF036035.1 | 4 | 1759 | AAD04257.1 | |
AF040710.1 | 4 | 1679 | AAC70915.1 | |
AF502909.1 | AAM60775.1 | |||
AF502910.1 | AAM60776.1 | |||
AF502911.1 | AAM60777.1 | |||
AF502912.1 | AAM60778.1 | |||
BC005896.1 | 4 | 1649 | AAH05896.1 | |
BC012892.1 | 3 | 1514 | AAH12892.1 | |
DB319349.1 | 1 | 187 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2759148 | Chr.3:50161045 - 50333946 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv523560 | Chr.3:50134964 - 50314769 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
dgv8317n54 | Chr.3:50206077 - 50383123 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv834693 | Chr.3:50251400 - 50470533 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv834692 | Chr.3:50161052 - 50356033 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
dgv8316n54 | Chr.3:50115923 - 50299991 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv470561 | Chr.3:50121341 - 50547568 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv523662 | Chr.3:50107518 - 50547568 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv590255 | Chr.3:50210391 - 50399396 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv954855 | Chr.3:50152468 - 50439169 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)