Assay Details
Target Gene Details
Entrez Gene ID: | 3991 |
Gene Name: | lipase E, hormone sensitive type |
Gene Aliases: |
AOMS4, FPLD6, HSL, LHS |
Location: |
Chr.19:42401507-42427426 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LIPE | NM_005357.3 | NP_005348.2 | ||
XM_005258937.3 | XP_005258994.1 | |||
XM_005258938.4 | XP_005258995.1 | |||
XM_005258939.3 | XP_005258996.2 | |||
XM_005258940.3 | XP_005258997.1 | |||
XM_006723218.2 | XP_006723281.1 | |||
XM_017026810.1 | XP_016882299.1 | |||
AK292482.1 | ||||
BC070041.1 | AAH70041.1 | |||
U40002.1 | AAC50666.1 |
Target Gene Details
Entrez Gene ID: | 100996307 |
Gene Name: | LIPE antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.19:42397148-42652355 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LIPE-AS1 | NR_073180.1 | |||
AK311181.1 | ||||
DW455857.1 |
Target Gene Details
Entrez Gene ID: | 101930071 |
Gene Name: | uncharacterized LOC101930071 |
Gene Aliases: |
- |
Location: |
Chr.19:42397128-42408452 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LOC101930071 | NR_126041.1 | |||
BU566442.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv953584 | Chr.19:42387549 - 42412748 on Build GRCh38 | Deletion | LIPE LIPE-AS1 CNFN LOC101930071 |
nsv520239 | Chr.19:41853387 - 42426852 on Build GRCh38 | Loss | MIR4323 PRR19 LOC101930071 MIR8077 LIPE-AS1 GRIK5 TMEM145 LOC100505585 RABAC1 LIPE CD79A MEGF8 LOC390937 CNFN RPS19 LOC100505622 POU2F2 PAFAH1B3 ARHGEF1 ZNF574 ATP1A3 CIC MIR6797 ZNF526 ERF GSK3A DEDD2 |
nsv579632 | Chr.19:42400501 - 42408251 on Build GRCh38 | Gain | LIPE LIPE-AS1 LOC101930071 |
nsv526186 | Chr.19:42172329 - 42699355 on Build GRCh38 | Gain | LIPE MEGF8 CNFN PRR19 POU2F2 PAFAH1B3 LOC101930071 MIR8077 LIPE-AS1 CIC CEACAM8 CEACAM1 ZNF526 TMEM145 ERF CXCL17 GSK3A DEDD2 |
nsv522854 | Chr.19:41613927 - 42426852 on Build GRCh38 | Gain | MIR4323 CEACAM4 PRR19 LOC101930071 MIR8077 DMRTC2 LIPE-AS1 GRIK5 TMEM145 CEACAM3 LYPD4 LOC100505585 CEACAM7 CEACAM6 RABAC1 LIPE CD79A MEGF8 LOC390937 CNFN RPS19 LOC100505622 POU2F2 PAFAH1B3 ARHGEF1 ZNF574 ATP1A3 CIC MIR6797 ZNF526 ERF CEACAM5 GSK3A DEDD2 |
esv2422471 | Chr.19:42228989 - 43165333 on Build GRCh38 | Deletion | PSG10P LIPE PSG11 MEGF8 PSG1 CNFN PRR19 PAFAH1B3 PSG3 LOC101930071 PSG6 PSG2 MIR8077 LIPE-AS1 CIC PSG8 CEACAM8 CEACAM1 TMEM145 ERF CXCL17 LOC100289650 GSK3A PSG7 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |