Assay Details
Target Gene Details
Entrez Gene ID: | 79585 |
Gene Name: | coronin 7 |
Gene Aliases: |
0610011B16Rik, CRN7, POD1 |
Location: |
Chr.16:4354542-4416961 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CORO7 | NM_001201472.1 | 12 | 1377 | NP_001188401.1 |
NM_001201473.1 | 10 | 1176 | NP_001188402.1 | |
NM_024535.4 | 12 | 1431 | NP_078811.3 | |
AK025674.1 | 12 | 1040 | BAB15211.1 | |
AK094596.1 | 12 | 1050 | ||
AK097238.1 | 12 | 897 | ||
AK294045.1 | 12 | 1012 | ||
AK296723.1 | 12 | 1009 | ||
AK296807.1 | 10 | 842 | ||
AK303353.1 | 7 | 562 | ||
AK309327.1 | 12 | 1005 | ||
AK309763.1 | ||||
BC117289.1 | 12 | 1050 | ||
BC117291.1 | 12 | 1050 |
Target Gene Details
Entrez Gene ID: | 100529144 |
Gene Name: | CORO7-PAM16 readthrough |
Gene Aliases: |
- |
Location: |
Chr.16:4340251-4416961 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CORO7-PAM16 | NM_001201479.1 | 12 | 1431 | NP_001188408.1 |
AL833954.1 | 13 | 1451 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv471075 | Chr.16:4317729 - 4395326 on Build GRCh38 | Loss | CORO7 GLIS2-AS1 VASN GLIS2 PAM16 CORO7-PAM16 |
nsv1046986 | Chr.16:4049000 - 5555721 on Build GRCh38 | Gain | UBALD1 SEC14L5 NAGPA ROGDI PPL ADCY9 NAGPA-AS1 ALG1 PAM16 EEF2KMT CDIP1 C16orf71 MIR6769A CORO7 VASN ANKS3 MGRN1 TFAP4 RBFOX1 DNAJA3 SRL ZNF500 GLIS2-AS1 NUDT16L1 GLYR1 NMRAL1 SMIM22 SEPT12 HMOX2 GLIS2 C16orf96 LINC01569 C16orf89 LOC105371063 CORO7-PAM16 UBN1 |
nsv571274 | Chr.16:4309420 - 4441189 on Build GRCh38 | Gain | DNAJA3 CORO7 GLIS2-AS1 VASN GLIS2 PAM16 CORO7-PAM16 |
nsv527866 | Chr.16:4206462 - 4412618 on Build GRCh38 | Loss | CORO7 SRL GLIS2-AS1 VASN GLIS2 PAM16 LINC01569 LOC105371063 CORO7-PAM16 TFAP4 |
esv3584924 | Chr.16:4330628 - 4469258 on Build GRCh38 | Gain | DNAJA3 CORO7 VASN GLIS2 PAM16 NMRAL1 CORO7-PAM16 |
nsv520212 | Chr.16:4354619 - 4390827 on Build GRCh38 | Loss | CORO7 VASN CORO7-PAM16 |
esv23732 | Chr.16:4340813 - 4476244 on Build GRCh38 | Gain | DNAJA3 CORO7 HMOX2 VASN PAM16 NMRAL1 CORO7-PAM16 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |