Assay Details
Target Gene Details
Entrez Gene ID: | 55471 |
Gene Name: | NADH:ubiquinone oxidoreductase complex assembly factor 7 |
Gene Aliases: |
C2orf56, MidA, PRO1853 |
Location: |
Chr.2:37231631-37259052 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NDUFAF7 | NM_001083946.1 | NP_001077415.1 | ||
NM_144736.4 | 5 | 532 | NP_653337.1 | |
XM_005264410.1 | 5 | 512 | XP_005264467.1 | |
XM_005264411.1 | XP_005264468.1 | |||
XM_011532956.1 | 5 | 468 | XP_011531258.1 | |
AK294240.1 | 5 | 466 | ||
AK299015.1 | ||||
AK302150.1 | 4 | 402 | ||
AK303555.1 | 5 | 480 | ||
BC004548.2 | 5 | 497 | AAH04548.2 | |
BI334629.1 | ||||
BM172474.1 | ||||
BX538031.1 | CAD97976.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |