Assay Details
Target Gene Details
Entrez Gene ID: | 64689 |
Gene Name: | golgi reassembly stacking protein 1 |
Gene Aliases: |
GOLPH5, GRASP65, P65 |
Location: |
Chr.3:39096599-39108363 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GORASP1 | NM_001278789.1 | 7 | 2506 | NP_001265718.1 |
NM_001278790.1 | 6 | 2326 | NP_001265719.1 | |
NM_031899.3 | 9 | 3254 | NP_114105.1 | |
NR_103866.1 | 9 | 2750 | ||
NR_103867.1 | 9 | 2742 | ||
XM_006713301.3 | 9 | 3354 | XP_006713364.1 | |
XM_011534020.2 | 9 | 3264 | XP_011532322.1 | |
XM_011534021.2 | 8 | 3174 | XP_011532323.1 | |
XM_017007050.1 | 9 | 2440 | XP_016862539.1 | |
XM_017007051.1 | 8 | 2789 | XP_016862540.1 | |
XM_017007052.1 | 8 | 2385 | XP_016862541.1 | |
AJ409349.1 | 9 | 2530 | CAC35160.1 | |
AK001574.1 | 8 | 2772 | ||
AK057043.1 | 6 | 2064 | ||
AK091168.1 | BAC03598.1 | |||
AK124755.1 | 9 | 3254 | ||
BC008928.2 | 9 | 2471 | AAH08928.1 | |
BC075854.1 | 9 | 2540 | AAH75854.1 | |
BC103826.1 | 9 | 2472 | AAI03827.1 | |
BC103827.1 | 9 | 2472 | AAI03828.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |