Assay Details
Target Gene Details
Entrez Gene ID: | 29947 |
Gene Name: | DNA methyltransferase 3 like |
Gene Aliases: |
- |
Location: |
Chr.21:44246339-44262216 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DNMT3L | NM_013369.3 | 6 | 911 | NP_037501.2 |
NM_175867.2 | 6 | 911 | NP_787063.1 | |
AF194032.1 | 6 | 911 | AAF05812.1 | |
BC002560.2 | 6 | 585 | AAH02560.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv587741 | Chr.21:44248288 - 44263467 on Build GRCh38 | Loss | DNMT3L LOC105372833 |
nsv953645 | Chr.21:44156018 - 44264017 on Build GRCh38 | Deletion | DNMT3L ICOSLG LOC105372833 |
nsv587743 | Chr.21:44255994 - 44263467 on Build GRCh38 | Gain | DNMT3L |
esv2723638 | Chr.21:44249174 - 44593709 on Build GRCh38 | Deletion | LRRC3-AS1 LOC100505727 LRRC3 TSPEAR DNMT3L C21orf2 KRTAP10-6 KRTAP10-3 TRPM2-AS KRTAP10-5 TSPEAR-AS1 KRTAP10-1 PFKL AIRE KRTAP10-4 TSPEAR-AS2 KRTAP10-2 TRPM2 LOC105372833 |
nsv587740 | Chr.21:44209682 - 44429981 on Build GRCh38 | Loss | TRPM2-AS DNMT3L PFKL AIRE C21orf2 ICOSLG TRPM2 LOC105372833 |
esv33817 | Chr.21:43749531 - 44647625 on Build GRCh38 | Loss | LOC100505727 PDXK KRTAP10-7 LRRC3 TSPEAR C21orf2 AATBC TSPEAR-AS1 KRTAP10-1 AIRE TSPEAR-AS2 TRPM2 LRRC3-AS1 KRTAP10-11 CSTB DNMT3L AGPAT3 ICOSLG KRTAP10-6 KRTAP10-3 TRPM2-AS KRTAP10-10 KRTAP10-5 KRTAP10-9 PWP2 KRTAP10-8 PFKL KRTAP10-4 TRAPPC10 KRTAP10-2 LOC105372833 RRP1 C21orf33 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |