Assay Details
Target Gene Details
Entrez Gene ID: | 10202 |
Gene Name: | dehydrogenase/reductase 2 |
Gene Aliases: |
HEP27, SDR25C1 |
Location: |
Chr.14:23630115-23645639 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DHRS2 | NM_005794.3 | 9 | 1366 | NP_005785.1 |
NM_182908.4 | 9 | 1377 | NP_878912.1 | |
XM_005267249.1 | 9 | 1078 | XP_005267306.1 | |
XM_006720001.3 | 9 | 1041 | XP_006720064.1 | |
XM_011536338.1 | 8 | 1007 | XP_011534640.1 | |
XM_011536339.2 | 8 | 857 | XP_011534641.1 | |
AB209653.1 | 7 | 5008 | BAD92890.1 | |
AK000345.1 | 1 | 2285 | ||
AK092834.1 | 5 | 2777 | ||
AK222857.1 | 9 | 1366 | BAD96577.1 | |
AK309945.1 | 9 | 2853 | ||
BC002786.2 | 9 | 1368 | AAH02786.1 | |
BC007339.2 | 8 | 892 | AAH07339.2 | |
U31875.1 | 9 | 1352 | AAA82048.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |