Assay Details
Target Gene Details
Entrez Gene ID: | 388364 |
Gene Name: | transmembrane and immunoglobulin domain containing 1 |
Gene Aliases: |
TMIGD, UNQ9372 |
Location: |
Chr.17:30316329-30334076 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TMIGD1 | NM_001319942.1 | 3 | 251 | NP_001306871.1 |
NM_206832.2 | 3 | 251 | NP_996663.1 | |
XM_011524787.1 | 3 | 265 | XP_011523089.1 | |
AK172838.1 | 3 | 251 | BAD18803.1 | |
AK290266.1 | 3 | 224 | ||
AY358153.1 | 3 | 239 | AAQ88520.1 | |
BC137201.1 | 3 | 235 | ||
BC137202.1 | 3 | 235 |