Assay Details
Target Gene Details
Entrez Gene ID: | 3029 |
Gene Name: | hydroxyacylglutathione hydrolase |
Gene Aliases: |
GLO2, GLX2, GLXII, HAGH1 |
Location: |
Chr.16:1809099-1827194 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
HAGH | NM_001040427.1 | 1 | 294 | NP_001035517.1 |
NM_001286249.1 | 1 | 294 | NP_001273178.1 | |
NM_005326.4 | 1 | 294 | NP_005317.2 | |
BC002627.2 | 1 | 240 | ||
BG703401.1 | 1 | 221 | ||
BI758286.1 | 1 | 261 | ||
DR006636.1 | 1 | 279 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv517196 | Chr.16:1729694 - 1844960 on Build GRCh38 | Gain+Loss | MAPK8IP3 MIR3177 NUBP2 MRPS34 EME2 FAHD1 MEIOB IGFALS NME3 HAGH SPSB3 |
nsv571175 | Chr.16:1806439 - 1875278 on Build GRCh38 | Loss | FAHD1 MEIOB HAGH |
nsv1040440 | Chr.16:1811739 - 1866102 on Build GRCh38 | Gain | FAHD1 MEIOB HAGH |
esv2422427 | Chr.16:10001 - 2723164 on Build GRCh38 | Duplication | AMDHD2 MIR3177 RPUSD1 ABCA3 WASIR2 LUC7L NTN3 CCDC154 TPSAB1 SPSB3 MCRIP2 PGP TSR3 ZNF598 RPS2 C1QTNF8 SNORA10 MRPL28 MIR3180-5 CCDC78 POLR3K E4F1 MIR6767 RGS11 LINC00235 HBQ1 NPW FLJ42627 SSTR5 LMF1-AS1 MEIOB RHBDL1 SLC9A3R2 ATP6V0C CAPN15 JMJD8 FBXL16 PRSS29P NME3 NHLRC4 SYNGR3 MIR940 LMF1 RHOT2 CHTF18 MIR6768 AXIN1 GFER MIR4516 LOC100287175 PDIA2 SNORA78 TELO2 RAB26 HBA2 ARHGDIG MIR3176 MIR3178 TPSD1 TPSB2 DNASE1L2 SOX8 RHBDF1 LOC100134368 KCTD5 MIR6859-4 LOC105371045 TSC2 WFIKKN1 CRAMP1 PTX4 HBM WDR24 MLST8 SSTR5-AS1 UNKL PRR25 C16orf59 SNRNP25 RAB11FIP3 LINC00254 TBL3 MIR3677 HN1L HAGHL FAM234A FAM173A UBE2I TRAF7 NOXO1 TMEM204 CASKIN1 ABCA17P BAIAP3 HBZ DDX11L10 RNF151 LOC105371038 WDR90 NTHL1 CLCN7 RPL3L LOC652276 MRPS34 GNG13 SNORD60 LOC107987233 MSRB1 LOC105371184 NME4 TBC1D24 PIGQ MPG PRSS27 HBA1 ECI1 EME2 IGFALS LOC106660606 LOC105371046 C16orf91 TPSG1 TMEM8A PRR35 MIR6511B1 LOC729652 HS3ST6 MSLN LOC107984876 SNHG9 NARFL CACNA1H PDPK1 ERVK13-1 RAB40C FAHD1 GNPTG LOC105371049 CEMP1 METRN MAPK8IP3 MIR1225 LOC101929280 IFT140 RNPS1 HAGH MIR4717 MIR662 SNORA64 RPL23AP5 NDUFB10 NUBP2 SNHG19 NPRL3 BRICD5 STUB1 PKD1 CCNF METTL26 LOC105371047 MIR5587 DECR2 |
nsv1051692 | Chr.16:1505070 - 1866102 on Build GRCh38 | Loss | MAPK8IP3 MIR3177 MRPS34 MEIOB HN1L IFT140 HAGH SPSB3 TELO2 CRAMP1 NUBP2 TMEM204 EME2 FAHD1 IGFALS NME3 LOC105371047 LOC105371046 |
nsv1042986 | Chr.16:1784723 - 1866102 on Build GRCh38 | Loss | NUBP2 FAHD1 MEIOB IGFALS HAGH |
nsv518641 | Chr.16:1645775 - 1835631 on Build GRCh38 | Loss | MAPK8IP3 MIR3177 MRPS34 MEIOB HN1L HAGH SPSB3 CRAMP1 NUBP2 EME2 FAHD1 IGFALS NME3 |
nsv952900 | Chr.16:1724500 - 1827999 on Build GRCh38 | Deletion | MAPK8IP3 MIR3177 NUBP2 MRPS34 EME2 FAHD1 IGFALS NME3 HAGH SPSB3 |
nsv457337 | Chr.16:1813519 - 1855005 on Build GRCh38 | Loss | FAHD1 MEIOB HAGH |
nsv571177 | Chr.16:1817039 - 1897015 on Build GRCh38 | Gain | FAHD1 LINC00254 MEIOB HAGH |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |