Assay Details
Target Gene Details
Entrez Gene ID: | 10272 |
Gene Name: | follistatin like 3 |
Gene Aliases: |
FLRG, FSRP |
Location: |
Chr.19:676389-683392 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
FSTL3 | NM_005860.2 | 5 | 1746 | NP_005851.1 |
AY358917.1 | 5 | 1727 | AAQ89276.1 | |
BC005839.2 | 5 | 1732 | AAH05839.1 | |
U76702.1 | 5 | 1718 | AAC64321.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv134n111 | Chr.19:607984 - 967584 on Build GRCh38 | Deletion | AZU1 PALM ARID3A RNU6-9 LOC101928450 MED16 MIR4745 MIR3187 FSTL3 PRSS57 ELANE RNF126 FGF22 PTBP1 PRTN3 POLRMT HCN2 R3HDM4 KISS1R MISP CFD PLPPR3 |
nsv428360 | Chr.19:232069 - 702131 on Build GRCh38 | Gain | SHC2 MADCAM1 PLPP2 TPGS1 C2CD4C CDC34 FSTL3 PRSS57 RNF126 FGF22 LOC101929060 GZMM MIER2 POLRMT HCN2 BSG ODF3L2 THEG |
nsv577978 | Chr.19:294526 - 782854 on Build GRCh38 | Gain | SHC2 PALM LOC101928450 MADCAM1 TPGS1 C2CD4C CDC34 FSTL3 PRSS57 RNF126 FGF22 LOC101929060 GZMM MIER2 POLRMT HCN2 MISP BSG ODF3L2 THEG |
nsv953938 | Chr.19:355801 - 1104501 on Build GRCh38 | Deletion | PALM RNU6-9 ABCA7 MIR4745 MIR3187 CNN2 TPGS1 FSTL3 PRSS57 TMEM259 GZMM GRIN3B HCN2 R3HDM4 MISP CFD PLPPR3 ODF3L2 THEG AZU1 SHC2 ARID3A LOC101928450 MADCAM1 MED16 RNU6-2 C2CD4C CDC34 ARHGAP45 POLR2E ELANE RNF126 FGF22 GPX4 PTBP1 WDR18 LOC105372235 PRTN3 POLRMT KISS1R BSG |
nsv470101 | Chr.19:401714 - 1008505 on Build GRCh38 | Loss | PALM RNU6-9 MIR4745 MIR3187 TPGS1 FSTL3 PRSS57 GZMM GRIN3B HCN2 R3HDM4 MISP CFD PLPPR3 ODF3L2 AZU1 SHC2 ARID3A LOC101928450 MADCAM1 MED16 C2CD4C CDC34 ELANE RNF126 FGF22 PTBP1 WDR18 LOC105372235 PRTN3 POLRMT KISS1R BSG |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |