Assay Details
Target Gene Details
Entrez Gene ID: | 8412 |
Gene Name: | breast cancer anti-estrogen resistance 3 |
Gene Aliases: |
NSP2, SH2D3B |
Location: |
Chr.1:93561741-93847150 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
BCAR3 | NM_001261408.1 | NP_001248337.1 | ||
NM_001261409.1 | NP_001248338.1 | |||
NM_001261410.1 | NP_001248339.1 | |||
NM_003567.3 | NP_003558.1 | |||
XM_011542251.2 | XP_011540553.1 | |||
XM_017002480.1 | XP_016857969.1 | |||
AF124250.1 | AAD28245.1 | |||
AK055442.1 | ||||
AL833121.1 | ||||
BC039895.1 | AAH39895.1 | |||
CN337133.1 | ||||
DA057195.1 | ||||
U92715.1 | AAC39777.1 |
Target Gene Details
Entrez Gene ID: | 100129046 |
Gene Name: | uncharacterized LOC100129046 |
Gene Aliases: |
- |
Location: |
Chr.1:93591968-93600030 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LOC100129046 | NR_034091.1 | 1 | 428 | |
AK307291.1 | 1 | 428 |
More Information
Set Membership: |
Intragenic Exonic Non-exonic non-DGV Variation |