Assay Details
Target Gene Details
Entrez Gene ID: | 81544 |
Gene Name: | glycerophosphodiester phosphodiesterase domain containing 5 |
Gene Aliases: |
GDE2, PP1665 |
Location: |
Chr.11:75434640-75525916 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 14 - Exon 15 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GDPD5 | NM_030792.6 | NP_110419.5 | ||
XM_006718697.3 | XP_006718760.1 | |||
XM_011545276.2 | XP_011543578.1 | |||
XM_011545277.2 | XP_011543579.1 | |||
XM_011545278.2 | XP_011543580.1 | |||
XM_011545279.2 | XP_011543581.1 | |||
XM_011545280.1 | XP_011543582.1 | |||
XM_011545281.2 | XP_011543583.1 | |||
XM_011545285.2 | XP_011543587.1 | |||
XM_011545286.1 | XP_011543588.1 | |||
XM_011545287.1 | XP_011543589.1 | |||
AF318351.1 | AAL55858.1 | |||
AF318377.1 | AAL55884.1 | |||
AF451987.1 | AAP97686.1 | |||
AK074848.1 | BAC11242.1 | |||
AK098808.1 | BAC05418.1 | |||
AK310490.1 | ||||
AL833941.1 | CAD38796.2 | |||
AY358477.1 | AAQ88841.1 | |||
BC018771.1 | AAH18771.1 | |||
BC033391.1 | AAH33391.1 |
More Information
Set Membership: |
Intragenic Non-exonic non-DGV Variation |