Assay Details
Target Gene Details
Entrez Gene ID: | 7075 |
Gene Name: | tyrosine kinase with immunoglobulin like and EGF like domains 1 |
Gene Aliases: |
JTK14, TIE |
Location: |
Chr.1:43300895-43323110 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 12 - Intron 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TIE1 | NM_001253357.1 | NP_001240286.1 | ||
NM_005424.4 | NP_005415.1 | |||
XM_005271163.1 | XP_005271220.1 | |||
XM_006710869.1 | XP_006710932.1 | |||
XM_017002207.1 | XP_016857696.1 | |||
AF217976.1 | AAG17219.1 | |||
AK225560.1 | ||||
AK296735.1 | ||||
AK300399.1 | ||||
AK300623.1 | ||||
AK307639.1 | ||||
BC038239.1 | AAH38239.1 | |||
EU826590.1 | ||||
X60957.1 | CAA43290.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv546128 | Chr.1:43301467 - 43451738 on Build GRCh38 | Loss | ELOVL1 CDC20 SZT2 MPL MIR6734 MED8 HYI MIR6735 TIE1 |
nsv829693 | Chr.1:43129457 - 43312977 on Build GRCh38 | Loss | C1orf210 FAM183A TMEM125 MIR6733 EBNA1BP2 TIE1 CFAP57 |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |