Assay Details
Target Gene Details
Entrez Gene ID: | 55683 |
Gene Name: | KAT8 regulatory NSL complex subunit 3 |
Gene Aliases: |
KIAA1310, NSL3, Rcd1 |
Location: |
Chr.2:96593155-96638379 on Build GRCh38 |
Assay Gene Location: | Within Exon 27 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
KANSL3 | NM_001115016.2 | 21 | 4696 | NP_001108488.1 |
NR_047653.1 | 21 | 4757 | ||
NR_047654.1 | 21 | 4751 | ||
NR_047655.1 | 21 | 4683 | ||
NR_047656.1 | 20 | 4605 | ||
NR_047657.1 | 21 | 4551 | ||
NR_047658.1 | 20 | 4450 | ||
XM_005263984.1 | 22 | 4773 | XP_005264041.1 | |
XM_005263987.1 | 21 | 4689 | XP_005264044.1 | |
XM_017004470.1 | 22 | 4767 | XP_016859959.1 | |
XM_017004473.1 | 22 | 4786 | XP_016859962.1 | |
XM_017004474.1 | 22 | 4681 | XP_016859963.1 | |
AA526958.1 | ||||
AB037731.1 | 21 | 4503 | BAA92548.1 | |
AF311326.1 | 23 | 4503 | AAG33852.1 | |
AK000943.1 | BAA91437.1 | |||
AK023813.1 | 21 | 4718 | BAB14688.1 | |
AK074379.1 | 2 | 2206 | ||
AY050169.1 | 21 | 4724 | AAL13159.1 | |
BC051763.1 | 20 | 4405 | AAH51763.1 | |
BC063792.1 | 21 | 4626 | AAH63792.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2720413 | Chr.2:96472591 - 96793459 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv469775 | Chr.2:96575189 - 96746754 on Build GRCh38 | Loss |
![]() ![]() ![]() |
nsv834306 | Chr.2:96575378 - 96747201 on Build GRCh38 | Loss |
![]() ![]() ![]() |
dgv607e201 | Chr.2:95855884 - 97561622 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv1135873 | Chr.2:95981454 - 97579645 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
esv3425717 | Chr.2:95859900 - 97515905 on Build GRCh38 | Duplication |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)