Assay Details
Target Gene Details
Entrez Gene ID: | 64478 |
Gene Name: | CUB and Sushi multiple domains 1 |
Gene Aliases: |
PPP1R24 |
Location: |
Chr.8:2935353-4995035 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 63 - Exon 64 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CSMD1 | NM_033225.5 | NP_150094.5 | ||
XM_011534752.2 | XP_011533054.1 | |||
XM_011534753.2 | XP_011533055.1 | |||
XM_011534754.1 | XP_011533056.1 | |||
XM_017013731.1 | XP_016869220.1 | |||
AB209502.1 | BAD92739.1 | |||
AF333704.2 | AAK73475.2 | |||
AK127129.1 | ||||
AY017307.1 | AAG52948.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1033589 | Chr.8:2490387 - 3447578 on Build GRCh38 | Loss |
![]() ![]() ![]() |
dgv1357e214 | Chr.8:2895602 - 3065852 on Build GRCh38 | Gain |
![]() ![]() |
esv3584570 | Chr.8:2827824 - 3129358 on Build GRCh38 | Gain |
![]() ![]() |
esv3891261 | Chr.8:2316229 - 3462226 on Build GRCh38 | Gain |
![]() ![]() ![]() |
dgv6815n100 | Chr.8:2193919 - 3295843 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() |
nsv609710 | Chr.8:2872547 - 3175339 on Build GRCh38 | Gain |
![]() ![]() |
dgv11848n54 | Chr.8:2320544 - 3445528 on Build GRCh38 | Gain |
![]() ![]() ![]() |
esv3615898 | Chr.8:2952268 - 2969870 on Build GRCh38 | Gain |
![]() |
esv3584572 | Chr.8:2935217 - 3102350 on Build GRCh38 | Gain |
![]() ![]() |
nsv1027569 | Chr.8:2660367 - 3086961 on Build GRCh38 | Loss |
![]() ![]() ![]() |
nsv1024325 | Chr.8:651043 - 3203837 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv517808 | Chr.8:2269008 - 3111255 on Build GRCh38 | Gain |
![]() ![]() ![]() |
nsv520737 | Chr.8:2951975 - 2967728 on Build GRCh38 | Gain |
![]() |
nsv1034294 | Chr.8:2901778 - 3054246 on Build GRCh38 | Gain |
![]() ![]() |
nsv1018407 | Chr.8:2866772 - 3447578 on Build GRCh38 | Gain |
![]() ![]() |
dgv6827n100 | Chr.8:2473547 - 3062661 on Build GRCh38 | Gain |
![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)