Assay Details
Target Gene Details
Entrez Gene ID: | 8507 |
Gene Name: | ectodermal-neural cortex 1 |
Gene Aliases: |
CCL28, ENC-1, KLHL35, KLHL37, NRPB, PIG10, TP53I10 |
Location: |
Chr.5:74627406-74641424 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ENC1 | NM_001256574.1 | 1 | 1063 | NP_001243503.1 |
NM_001256575.1 | 1 | 1063 | NP_001243504.1 | |
NM_001256576.1 | 1 | 1063 | NP_001243505.1 | |
NM_003633.3 | 1 | 1063 | NP_003624.1 | |
NR_046318.1 | 1 | 1063 | ||
XM_011543696.2 | 1 | 1063 | XP_011541998.1 | |
XM_011543697.2 | 1 | 1063 | XP_011541999.1 | |
AF005381.1 | AAC64498.1 | |||
AF010314.1 | 1 | 353 | AAC39532.1 | |
AF059611.1 | 1 | 331 | AAC26109.1 | |
AK096578.1 | 1 | 1063 | ||
AK295128.1 | 1 | 368 | ||
AK299624.1 | ||||
AK312608.1 | 1 | 365 | ||
BC000418.2 | 1 | 333 | AAH00418.1 | |
DA032687.1 | 1 | 365 | ||
DA498140.1 | 1 | 365 | ||
DA785554.1 | 1 | 365 |
Target Gene Details
Entrez Gene ID: | 3074 |
Gene Name: | hexosaminidase subunit beta |
Gene Aliases: |
ENC-1AS, HEL-248, HEL-S-111 |
Location: |
Chr.5:74640023-74721288 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
HEXB | NM_001292004.1 | 1 | 340 | NP_001278933.1 |
AY643499.1 | 1 | 345 | AAV48875.1 | |
FJ224336.1 | 1 | 345 | ||
GQ472222.1 | 1 | 345 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |