Assay Details
Target Gene Details
Entrez Gene ID: | 5818 |
Gene Name: | nectin cell adhesion molecule 1 |
Gene Aliases: |
CD111, CLPED1, ED4, HIgR, HV1S, HVEC, OFC7, PRR, PRR1, PVRL1, PVRR, PVRR1, SK-12, nectin-1 |
Location: |
Chr.11:119638098-119728725 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NECTIN1 | NM_203285.1 | NP_976030.1 | ||
AF110314.1 | AAD04944.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv509445 | Chr.11:119609244 - 119695172 on Build GRCh38 | Insertion |
![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)