Assay Details
Target Gene Details
Entrez Gene ID: | 201501 |
Gene Name: | zinc finger and BTB domain containing 7C |
Gene Aliases: |
APM-1, APM1, ZBTB36, ZNF857C |
Location: |
Chr.18:48026886-48412639 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ZBTB7C | NM_001039360.2 | 3 | 3871 | NP_001034449.1 |
NM_001318841.1 | 5 | 4052 | NP_001305770.1 | |
XM_005258229.4 | 4 | 3796 | XP_005258286.1 | |
XM_011525861.2 | 7 | 4276 | XP_011524163.1 | |
XM_011525863.2 | 6 | 4393 | XP_011524165.1 | |
XM_011525864.2 | 6 | 4148 | XP_011524166.1 | |
XM_011525865.2 | 6 | 4389 | XP_011524167.1 | |
XM_011525866.2 | 5 | 4134 | XP_011524168.1 | |
XM_011525867.2 | 5 | 3923 | XP_011524169.1 | |
XM_011525869.2 | 6 | 4398 | XP_011524171.1 | |
XM_011525870.2 | 5 | 4094 | XP_011524172.1 | |
XM_011525871.2 | 3 | 4609 | XP_011524173.1 | |
XM_017025605.1 | 7 | 6071 | XP_016881094.1 | |
XM_017025606.1 | 7 | 4525 | XP_016881095.1 | |
XM_017025607.1 | 7 | 4280 | XP_016881096.1 | |
XM_017025608.1 | 6 | 5948 | XP_016881097.1 | |
XM_017025609.1 | 5 | 4265 | XP_016881098.1 | |
AK095226.1 | 1 | 1608 | ||
AM988836.1 | 3 | 3859 | ||
AM988837.1 | 3 | 4004 | ||
AM988838.1 | 4 | 4054 | ||
AM988839.1 | 5 | 4052 | ||
BM983365.1 | 1 | 318 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1066479 | Chr.18:47979198 - 48028136 on Build GRCh38 | Gain | ZBTB7C |
esv274948 | Chr.18:48025919 - 48031491 on Build GRCh38 | Loss | ZBTB7C |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |