Assay Details
Target Gene Details
Entrez Gene ID: | 60343 |
Gene Name: | family with sequence similarity 3 member A |
Gene Aliases: |
2.19, DLD, DXS560S, XAP-7 |
Location: |
Chr.X:154506159-154516249 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 7 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv33784 | Chr.X:153962665 - 155699588 on Build GRCh38 | Gain+Loss | RAB39B LOC105373386 OPN1MW2 FLNA TMLHE-AS1 TAZ HCFC1 LOC101927830 OPN1LW MPP1 FAM50A CTAG1B OPN1MW DKC1 SNORA70 FAM223B UBL4A MTCP1 TEX28 SLC10A3 MIR1184-2 F8A3 H2AFB1 HCFC1-AS1 F8A2 CMC4 SPRY3 VBP1 G6PD MIR1184-1 TKTL1 IRAK1 MIR3202-1 FUNDC2 ATP6AP1 LOC107987332 F8A1 H2AFB2 CH17-340M24.3 GAB3 CTAG2 OPN1MW3 MIR664B H2AFB3 CLIC2 MIR3202-2 TMLHE MECP2 PLXNA3 FAM223A BRCC3 LAGE3 GDI1 SNORA36A MIR6858 F8 MIR718 IKBKG CTAG1A SNORA56 DNASE1L1 FAM3A MIR1184-3 SMIM9 RPL10 TMEM187 EMD |
nsv519042 | Chr.X:154082978 - 154601444 on Build GRCh38 | Gain | LOC105373386 OPN1MW2 FLNA TAZ ATP6AP1 LOC107987332 OPN1LW FAM50A CH17-340M24.3 OPN1MW OPN1MW3 SNORA70 UBL4A MECP2 PLXNA3 TEX28 FAM223A LAGE3 GDI1 MIR6858 SLC10A3 IKBKG CTAG1A DNASE1L1 FAM3A RPL10 EMD G6PD TKTL1 |
esv33199 | Chr.X:153065734 - 154591978 on Build GRCh38 | Gain+Loss | LOC105373386 OPN1MW2 AVPR2 FLNA MAGEA1 TAZ ZNF275 ZFP92 HCFC1 OPN1LW FAM50A PNMA6A OPN1MW SNORA70 LOC105373378 IDH3G UBL4A BGN TEX28 ARHGAP4 DUSP9 SLC10A3 LOC105373383 HCFC1-AS1 FAM58A LOC107983954 L1CAM G6PD TKTL1 IRAK1 HAUS7 MIR3202-1 LOC105373381 PDZD4 ATP6AP1 LOC107987332 PLXNB3 CH17-340M24.3 SRPK3 OPN1MW3 NAA10 MIR3202-2 MECP2 LCA10 PLXNA3 FAM223A BCAP31 LAGE3 GDI1 MIR6858 TREX2 MIR718 IKBKG ATP2B3 ABCD1 CTAG1A DNASE1L1 LOC105373377 FAM3A RPL10 PNCK SSR4 TMEM187 SLC6A8 EMD RENBP |
nsv508816 | Chr.X:154492171 - 154591783 on Build GRCh38 | Insertion | FAM223A FAM3A IKBKG G6PD CTAG1A |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |