Assay Details
Target Gene Details
Entrez Gene ID: | 57591 |
Gene Name: | megakaryoblastic leukemia (translocation) 1 |
Gene Aliases: |
BSAC, MAL, MRTF-A |
Location: |
Chr.22:40410281-40636719 on Build GRCh38 |
Assay Gene Location: | Within Exon 19 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MKL1 | NM_001282660.1 | 12 | 2956 | NP_001269589.1 |
NM_001282661.1 | 14 | 3218 | NP_001269590.1 | |
NM_001282662.1 | 15 | 3378 | NP_001269591.1 | |
NM_001318139.1 | 13 | 3093 | NP_001305068.1 | |
NM_020831.4 | 15 | 3368 | NP_065882.1 | |
XM_005261692.1 | 14 | 3173 | XP_005261749.1 | |
XM_005261694.1 | 14 | 3263 | XP_005261751.1 | |
XM_011530283.1 | 16 | 3476 | XP_011528585.1 | |
XM_011530285.1 | 16 | 3387 | XP_011528587.1 | |
XM_011530287.1 | 8 | 2355 | XP_011528589.1 | |
XM_017028888.1 | 15 | 3451 | XP_016884377.1 | |
XM_017028889.1 | 11 | 4451 | XP_016884378.1 | |
AB037859.2 | 15 | 3340 | ||
AF368061.1 | 15 | 3293 | AAK54721.1 | |
AJ297257.1 | 12 | 2950 | CAC38826.1 | |
AJ297258.1 | 15 | 3350 | CAC38827.1 | |
AL713710.1 | 7 | 2163 | CAD28507.2 | |
BC114364.2 | 2 | 1543 | AAI14365.1 | |
BC115039.1 | 15 | 3344 | ||
CR456522.1 | 12 | 2767 | CAG30408.1 | |
HG764171.1 | 13 | 3093 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv829233 | Chr.22:40404227 - 40438253 on Build GRCh38 | Loss | SGSM3 MKL1 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |