Assay Details
Target Gene Details
Entrez Gene ID: | 6346 |
Gene Name: | C-C motif chemokine ligand 1 |
Gene Aliases: |
I-309, P500, SCYA1, SISe, TCA3 |
Location: |
Chr.17:34360328-34363233 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CCL1 | NM_002981.2 | NP_002972.1 | ||
AK312183.1 | ||||
BC105073.1 | AAI05074.1 | |||
BC105075.1 | AAI05076.1 | |||
DA620119.1 | ||||
M57502.1 | AAA61196.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv3141n100 | Chr.17:33630132 - 34611003 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv517423 | Chr.17:33777656 - 34498242 on Build GRCh38 | Gain+Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv574714 | Chr.17:33640441 - 34608994 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
esv3572553 | Chr.17:33632208 - 34617133 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv519297 | Chr.17:33640660 - 34608994 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)