Assay Details
Target Gene Details
Entrez Gene ID: | 116444 |
Gene Name: | glutamate ionotropic receptor NMDA type subunit 3B |
Gene Aliases: |
GluN3B, NR3B |
Location: |
Chr.19:1000419-1009814 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GRIN3B | NM_138690.2 | NP_619635.1 | ||
XM_017026243.1 | XP_016881732.1 | |||
AY507106.1 | AAS87020.1 | |||
AY507107.1 | AAS87021.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv9662 | Chr.19:893394 - 1317740 on Build GRCh38 | Gain | GPX4 ABCA7 CBARP GRIN3B KISS1R TMEM259 R3HDM4 ATP5D WDR18 POLR2E EFNA2 RNU6-2 STK11 LOC105372235 CIRBP-AS1 ARHGAP45 MIDN SBNO2 LOC102725180 RNU6-9 C19orf24 ARID3A CNN2 CIRBP |
nsv1160555 | Chr.19:887275 - 1346084 on Build GRCh38 | Deletion | GPX4 R3HDM4 ATP5D POLR2E EFNA2 RNU6-2 MED16 CIRBP-AS1 MIDN LOC102725180 RNU6-9 ARID3A CNN2 ABCA7 CBARP GRIN3B KISS1R TMEM259 WDR18 STK11 LOC105372235 ARHGAP45 SBNO2 C19orf24 CIRBP |
nsv578188 | Chr.19:923744 - 1059005 on Build GRCh38 | Loss | LOC105372235 ABCA7 GRIN3B TMEM259 WDR18 RNU6-2 ARID3A CNN2 |
nsv469845 | Chr.19:951640 - 1144486 on Build GRCh38 | Loss | GPX4 LOC105372235 ABCA7 ARHGAP45 GRIN3B TMEM259 SBNO2 WDR18 POLR2E RNU6-2 ARID3A CNN2 |
esv2758743 | Chr.19:951641 - 1544934 on Build GRCh38 | Gain+Loss | GPX4 NDUFS7 ADAMTSL5 DAZAP1 MUM1 ATP5D POLR2E EFNA2 RNU6-2 CIRBP-AS1 MIDN GAMT LOC102725180 ARID3A CNN2 ABCA7 CBARP GRIN3B REEP6 TMEM259 RPS15 WDR18 APC2 STK11 LOC105372235 PCSK4 ARHGAP45 PLK5 C19orf25 SBNO2 C19orf24 CIRBP |
nsv953938 | Chr.19:355801 - 1104501 on Build GRCh38 | Deletion | GPX4 MIR4745 HCN2 PRTN3 R3HDM4 PALM MIR3187 POLR2E RNU6-2 MED16 PTBP1 RNF126 SHC2 AZU1 LOC101928450 CDC34 FSTL3 THEG MADCAM1 RNU6-9 PRSS57 ARID3A CNN2 BSG FGF22 ABCA7 GRIN3B ELANE KISS1R TMEM259 TPGS1 PLPPR3 POLRMT WDR18 GZMM LOC105372235 C2CD4C ARHGAP45 MISP CFD ODF3L2 |
nsv470101 | Chr.19:401714 - 1008505 on Build GRCh38 | Loss | MIR4745 HCN2 PRTN3 R3HDM4 PALM MIR3187 MED16 PTBP1 RNF126 SHC2 AZU1 LOC101928450 CDC34 FSTL3 MADCAM1 RNU6-9 PRSS57 ARID3A BSG FGF22 GRIN3B ELANE KISS1R TPGS1 PLPPR3 POLRMT WDR18 GZMM LOC105372235 C2CD4C MISP CFD ODF3L2 |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |