Assay Details
Target Gene Details
Entrez Gene ID: | 3029 |
Gene Name: | hydroxyacylglutathione hydrolase |
Gene Aliases: |
GLO2, GLX2, GLXII, HAGH1 |
Location: |
Chr.16:1809099-1827194 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
HAGH | NM_001040427.1 | 1 | 152 | NP_001035517.1 |
NM_001286249.1 | 1 | 152 | NP_001273178.1 | |
NM_005326.4 | 1 | 152 | NP_005317.2 | |
AY576804.1 | ||||
BC002627.2 | 1 | 98 | ||
BG703401.1 | 1 | 79 | ||
BI758286.1 | 1 | 119 | ||
DR006636.1 | 1 | 137 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv517196 | Chr.16:1729694 - 1844960 on Build GRCh38 | Gain+Loss | EME2 NUBP2 NME3 MEIOB IGFALS FAHD1 SPSB3 MAPK8IP3 MIR3177 MRPS34 HAGH |
nsv571175 | Chr.16:1806439 - 1875278 on Build GRCh38 | Loss | MEIOB FAHD1 HAGH |
nsv1040440 | Chr.16:1811739 - 1866102 on Build GRCh38 | Gain | MEIOB FAHD1 HAGH |
esv2422427 | Chr.16:10001 - 2723164 on Build GRCh38 | Duplication | WASIR2 PRSS27 NUBP2 POLR3K UBE2I CHTF18 PRSS29P KCTD5 SPSB3 MLST8 CCDC154 LOC106660606 CRAMP1 MIR4717 CAPN15 LOC101929280 GNPTG TMEM204 BRICD5 C1QTNF8 MRPS34 PIGQ TPSD1 PGP SSTR5 E4F1 PTX4 MIR940 SNORA78 ARHGDIG HAGH SOX8 LINC00235 METTL26 LOC107984876 WDR90 METRN MIR3177 FBXL16 BAIAP3 NPW STUB1 RPUSD1 HBZ SSTR5-AS1 RAB11FIP3 NPRL3 TSR3 FAM234A HS3ST6 LOC107987233 RHBDL1 LOC105371047 PDPK1 MSRB1 TRAF7 NME4 RPL23AP5 PDIA2 MIR3176 AXIN1 MIR1225 WFIKKN1 ATP6V0C ECI1 LMF1 RGS11 LOC652276 TBC1D24 EME2 NDUFB10 RNF151 MIR6767 FAHD1 PRR35 ABCA17P MIR5587 SYNGR3 HBA1 NME3 C16orf91 HBM LOC105371049 MEIOB MPG MSLN LOC105371045 CASKIN1 WDR24 SNRNP25 JMJD8 MIR3677 TPSG1 CCNF RAB40C IFT140 LMF1-AS1 GNG13 NHLRC4 SNHG19 SNORD60 CEMP1 LOC105371038 RPL3L MIR662 MAPK8IP3 SNHG9 TELO2 MIR4516 FLJ42627 DECR2 TPSB2 TPSAB1 UNKL CCDC78 PKD1 NARFL TMEM8A MIR6859-4 MIR6511B1 MRPL28 CLCN7 ABCA3 TSC2 RNPS1 NTHL1 HAGHL FAM173A SLC9A3R2 AMDHD2 DNASE1L2 RAB26 MIR6768 C16orf59 MIR3178 LINC00254 MCRIP2 PRR25 LOC729652 HBA2 SNORA64 LOC105371184 HN1L LUC7L HBQ1 CACNA1H SNORA10 NTN3 RHBDF1 GFER MIR3180-5 DDX11L10 TBL3 ERVK13-1 RPS2 LOC100287175 NOXO1 LOC100134368 IGFALS RHOT2 LOC105371046 ZNF598 |
nsv1051692 | Chr.16:1505070 - 1866102 on Build GRCh38 | Loss | NUBP2 NME3 HN1L MEIOB SPSB3 MAPK8IP3 TELO2 HAGH LOC105371047 EME2 CRAMP1 IGFALS FAHD1 IFT140 TMEM204 LOC105371046 MIR3177 MRPS34 |
nsv1042986 | Chr.16:1784723 - 1866102 on Build GRCh38 | Loss | NUBP2 MEIOB IGFALS FAHD1 HAGH |
nsv518641 | Chr.16:1645775 - 1835631 on Build GRCh38 | Loss | NUBP2 NME3 HN1L MEIOB SPSB3 MAPK8IP3 HAGH EME2 CRAMP1 IGFALS FAHD1 MIR3177 MRPS34 |
nsv457337 | Chr.16:1813519 - 1855005 on Build GRCh38 | Loss | MEIOB FAHD1 HAGH |
nsv952900 | Chr.16:1724500 - 1827999 on Build GRCh38 | Deletion | EME2 NUBP2 NME3 IGFALS FAHD1 SPSB3 MAPK8IP3 MIR3177 MRPS34 HAGH |
nsv571177 | Chr.16:1817039 - 1897015 on Build GRCh38 | Gain | MEIOB FAHD1 LINC00254 HAGH |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |