Assay Details
Target Gene Details
Entrez Gene ID: | 339229 |
Gene Name: | oxidoreductase like domain containing 1 |
Gene Aliases: |
C17orf90 |
Location: |
Chr.17:81665036-81666637 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
OXLD1 | NM_001039842.2 | 2 | 478 | NP_001034931.1 |
NM_001304994.1 | 2 | 832 | NP_001291923.1 | |
NM_001304995.1 | 2 | 719 | NP_001291924.1 | |
NM_001304999.1 | 1 | 1411 | NP_001291928.1 | |
NR_130936.1 | 2 | 841 | ||
NR_130937.1 | 2 | 846 | ||
NR_130938.1 | 2 | 850 | ||
NR_130939.1 | 2 | 507 | ||
AY007126.1 | 1 | 1409 | ||
BC090923.1 | 2 | 429 | AAH90923.1 | |
BU194770.1 | 2 | 677 | ||
CB048026.1 | 2 | 515 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv26223 | Chr.17:80659041 - 81736487 on Build GRCh38 | Gain+Loss | LOC100130370 BAHCC1 LOC100129503 MIR4740 C17orf89 MIR3065 TMEM105 HGS AATK-AS1 FSCN2 SLC25A10 AATK LOC101928855 MIR6786 MIR3186 MRPL12 TEPSIN PDE6G FAAP100 CEP131 TSPAN10 RPTOR LOC105371921 MIR657 LOC105371925 LOC107985021 OXLD1 LINC00482 MIR1250 LOC400627 CCDC137 SLC38A10 ACTG1 NPLOC4 ARL16 BAIAP2-AS1 CHMP6 MIR338 LOC107985022 BAIAP2 |
nsv428351 | Chr.17:81152631 - 83064073 on Build GRCh38 | Gain | LOC100130370 BAHCC1 DCXR ALYREF MIR4740 C17orf89 MYADML2 TMEM105 GCGR PYCR1 ANAPC11 FSCN2 PCYT2 SLC25A10 ARHGDIA AATK NARF MIR6786 MRPL12 ZNF750 TBCD RFNG LOC105371925 MIR6787 WDR45B OXLD1 LINC00482 SIRT7 STRA13 LOC105371944 SNORD134 CCDC137 SLC38A10 FOXK2 UTS2R LOC101929552 LOC105376790 SECTM1 TEX19 MAFG LOC101929511 NPB FN3KRP OGFOD3 CCDC57 MCRIP1 GPS1 DUS1L HGS AATK-AS1 MIR4525 RAC3 MIR3186 TEPSIN PDE6G FAAP100 NOTUM CEP131 TSPAN10 ASPSCR1 C17orf62 LRRC45 LOC105376791 RAB40B CD7 CSNK1D FASN ACTG1 NPLOC4 MAFG-AS1 ARL16 SLC16A3 B3GNTL1 P4HB PPP1R27 HEXDC FN3K |
nsv470620 | Chr.17:81516973 - 82064485 on Build GRCh38 | Loss | DCXR ALYREF MCRIP1 GPS1 DUS1L MYADML2 GCGR HGS PYCR1 ANAPC11 FSCN2 PCYT2 SLC25A10 ARHGDIA RAC3 MIR6786 MRPL12 PDE6G FAAP100 NOTUM TSPAN10 RFNG ASPSCR1 LRRC45 OXLD1 SIRT7 STRA13 CCDC137 NPLOC4 MAFG-AS1 ARL16 P4HB MAFG PPP1R27 NPB |
nsv952390 | Chr.17:81637175 - 81860124 on Build GRCh38 | Deletion | MCRIP1 OXLD1 GCGR HGS CCDC137 SLC25A10 MIR6786 ARL16 MRPL12 P4HB PDE6G PPP1R27 TSPAN10 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |