Assay Details
Target Gene Details
Entrez Gene ID: | 81889 |
Gene Name: | fumarylacetoacetate hydrolase domain containing 1 |
Gene Aliases: |
C16orf36, YISKL |
Location: |
Chr.16:1827224-1840207 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
FAHD1 | NM_001018104.2 | NP_001018114.1 | ||
NM_001142398.1 | NP_001135870.1 | |||
BC063017.1 | AAH63017.1 |
Target Gene Details
Entrez Gene ID: | 254528 |
Gene Name: | meiosis specific with OB domains |
Gene Aliases: |
C16orf73, gs129 |
Location: |
Chr.16:1833983-1872178 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 13 - Exon 14 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MEIOB | NM_001163560.2 | NP_001157032.1 | ||
NM_152764.2 | NP_689977.2 | |||
BC029829.1 | AAH29829.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv517196 | Chr.16:1729694 - 1844960 on Build GRCh38 | Gain+Loss | IGFALS EME2 NUBP2 MIR3177 FAHD1 MEIOB NME3 SPSB3 MRPS34 HAGH MAPK8IP3 |
nsv833119 | Chr.16:1834308 - 1911484 on Build GRCh38 | Loss | FAHD1 LINC00254 MEIOB HS3ST6 |
nsv571175 | Chr.16:1806439 - 1875278 on Build GRCh38 | Loss | FAHD1 MEIOB HAGH |
nsv1040440 | Chr.16:1811739 - 1866102 on Build GRCh38 | Gain | FAHD1 MEIOB HAGH |
esv2422427 | Chr.16:10001 - 2723164 on Build GRCh38 | Duplication | RHOT2 C1QTNF8 NHLRC4 CAPN15 MSRB1 SOX8 TSC2 LOC105371038 GNPTG LMF1-AS1 LOC652276 KCTD5 TPSG1 LOC105371184 TPSAB1 NME4 SSTR5-AS1 RNPS1 RPS2 ECI1 C16orf59 ARHGDIG HBM TRAF7 METRN LOC105371046 MIR940 NPRL3 MIR4516 RNF151 UBE2I WASIR2 MIR6859-4 CCDC78 MLST8 HBZ MPG SNHG9 CHTF18 SNORA78 MIR3677 RAB11FIP3 PIGQ LMF1 FLJ42627 AMDHD2 FAM173A ATP6V0C TSR3 IFT140 CLCN7 SNRNP25 PDIA2 HS3ST6 HBA2 SNHG19 PRSS29P NUBP2 SLC9A3R2 SYNGR3 MSLN METTL26 LOC107984876 MAPK8IP3 NME3 E4F1 LOC105371045 FBXL16 NDUFB10 PGP MIR1225 MCRIP2 POLR3K TBC1D24 RHBDF1 MIR3180-5 HAGH LOC729652 LINC00254 JMJD8 GFER MIR6768 TMEM8A LOC101929280 ABCA17P RPUSD1 LUC7L RPL3L DDX11L10 LOC107987233 BAIAP3 SNORA10 SNORA64 SPSB3 STUB1 PKD1 BRICD5 CEMP1 CRAMP1 MIR3177 WFIKKN1 TPSB2 FAM234A NTHL1 RHBDL1 HAGHL SNORD60 CACNA1H PDPK1 TELO2 HBA1 LOC105371047 MIR3178 RGS11 PRR25 MIR5587 ZNF598 WDR24 NARFL IGFALS MIR3176 CCNF CASKIN1 DECR2 RAB40C AXIN1 RAB26 MRPS34 RPL23AP5 ABCA3 LOC105371049 CCDC154 LOC100287175 WDR90 TPSD1 LINC00235 DNASE1L2 FAHD1 C16orf91 NOXO1 NTN3 PTX4 GNG13 TBL3 EME2 UNKL LOC100134368 SSTR5 LOC106660606 NPW MIR6767 TMEM204 PRSS27 MIR662 HBQ1 MIR6511B1 MEIOB PRR35 HN1L MRPL28 MIR4717 ERVK13-1 |
nsv1051692 | Chr.16:1505070 - 1866102 on Build GRCh38 | Loss | IGFALS EME2 LOC105371046 CRAMP1 MIR3177 NME3 TMEM204 IFT140 MRPS34 TELO2 NUBP2 FAHD1 LOC105371047 MEIOB HN1L SPSB3 HAGH MAPK8IP3 |
nsv1042986 | Chr.16:1784723 - 1866102 on Build GRCh38 | Loss | IGFALS NUBP2 FAHD1 MEIOB HAGH |
nsv518641 | Chr.16:1645775 - 1835631 on Build GRCh38 | Loss | IGFALS EME2 CRAMP1 MIR3177 NME3 MRPS34 NUBP2 FAHD1 MEIOB HN1L SPSB3 HAGH MAPK8IP3 |
nsv457337 | Chr.16:1813519 - 1855005 on Build GRCh38 | Loss | FAHD1 MEIOB HAGH |
nsv571177 | Chr.16:1817039 - 1897015 on Build GRCh38 | Gain | FAHD1 LINC00254 MEIOB HAGH |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |