Assay Details
Target Gene Details
Entrez Gene ID: | 29115 |
Gene Name: | SAP30 binding protein |
Gene Aliases: |
HCNGP, HTRG, HTRP |
Location: |
Chr.17:75667116-75708062 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv833544 | Chr.17:75595541 - 75771639 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv470603 | Chr.17:75468213 - 75741366 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv516133 | Chr.17:75420733 - 75763223 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv576027 | Chr.17:75249939 - 75750289 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv833543 | Chr.17:75517604 - 75692749 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)