Assay Details
Target Gene Details
Entrez Gene ID: | 7434 |
Gene Name: | vasoactive intestinal peptide receptor 2 |
Gene Aliases: |
C16DUPq36.3, DUP7q36.3, PACAP-R-3, PACAP-R3, VIP-R-2, VPAC2, VPAC2R, VPCAP2R |
Location: |
Chr.7:159028175-159144957 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 17 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
VIPR2 | NM_001304522.1 | NP_001291451.1 | ||
NM_001308259.1 | NP_001295188.1 | |||
NM_003382.4 | NP_003373.2 | |||
NR_130758.1 | ||||
XM_005249561.3 | XP_005249618.1 | |||
XM_017012580.1 | XP_016868069.1 | |||
CA449700.1 | ||||
X95097.2 | CAA64474.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv465262 | Chr.7:158891461 - 159127494 on Build GRCh38 | Gain | WDR60 VIPR2 LINC00689 |
esv3615741 | Chr.7:158774147 - 159247998 on Build GRCh38 | Gain | WDR60 VIPR2 ESYT2 LOC105379752 LINC00689 |
nsv951684 | Chr.7:159006110 - 159037209 on Build GRCh38 | Deletion | VIPR2 LINC00689 |
dgv11804n54 | Chr.7:158818315 - 159127494 on Build GRCh38 | Gain | WDR60 VIPR2 ESYT2 LINC00689 |
dgv6772n100 | Chr.7:158645675 - 159162536 on Build GRCh38 | Gain | NCAPG2 WDR60 VIPR2 ESYT2 LOC105379752 LINC00689 |
nsv609419 | Chr.7:158712157 - 159117938 on Build GRCh38 | Gain | WDR60 VIPR2 ESYT2 LOC105379752 LINC00689 |
nsv609363 | Chr.7:158638215 - 159135878 on Build GRCh38 | Loss | NCAPG2 WDR60 VIPR2 ESYT2 LOC105379752 LINC00689 |
nsv526756 | Chr.7:158918188 - 159326796 on Build GRCh38 | Gain | WDR60 VIPR2 LINC00689 |
nsv1027593 | Chr.7:159014422 - 159131365 on Build GRCh38 | Loss | VIPR2 LINC00689 |
nsv1027282 | Chr.7:158995838 - 159162536 on Build GRCh38 | Gain | VIPR2 LINC00689 |
nsv519749 | Chr.7:158875884 - 159257090 on Build GRCh38 | Loss | WDR60 VIPR2 LINC00689 |
nsv1032688 | Chr.7:158869559 - 159327018 on Build GRCh38 | Loss | WDR60 VIPR2 LINC00689 |
esv3576497 | Chr.7:158778556 - 159091127 on Build GRCh38 | Gain | WDR60 VIPR2 ESYT2 LOC105379752 LINC00689 |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |