Assay Details
Target Gene Details
Entrez Gene ID: | 57611 |
Gene Name: | immunoglobulin superfamily containing leucine rich repeat 2 |
Gene Aliases: |
LINX |
Location: |
Chr.15:74128203-74141826 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ISLR2 | NM_001130136.1 | 4 | 1999 | NP_001123608.1 |
NM_001130137.1 | 4 | 1735 | NP_001123609.1 | |
NM_001130138.1 | 4 | 1443 | NP_001123610.1 | |
NM_020851.2 | 3 | 1568 | NP_065902.1 | |
XM_011521840.2 | 4 | 1838 | XP_011520142.1 | |
XM_011521841.1 | 4 | 1521 | XP_011520143.1 | |
XM_017022446.1 | 3 | 1670 | XP_016877935.1 | |
AB040898.2 | 4 | 1999 | ||
AK092297.1 | 3 | 1394 | ||
AK092302.1 | 4 | 2015 | ||
AK096602.1 | 2 | 1346 | ||
AK290467.1 | 3 | 1437 | ||
AY358316.1 | 3 | 1366 | AAQ88682.1 | |
BC114551.1 | AAI14552.1 | |||
BC152429.1 | 4 | 1999 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv833053 | Chr.15:74067815 - 74262799 on Build GRCh38 | Loss | STRA6 ISLR2 LOC283731 ISLR LOC732265 CCDC33 GOLGA6A |
nsv1050884 | Chr.15:74102642 - 74197072 on Build GRCh38 | Gain | STRA6 ISLR2 LOC283731 ISLR |
nsv469619 | Chr.15:73999518 - 74173980 on Build GRCh38 | Loss | ISLR2 LOC283731 ISLR LOC732265 GOLGA6A PML |
nsv827385 | Chr.15:74127198 - 74136960 on Build GRCh38 | Gain | ISLR2 LOC283731 |
nsv951993 | Chr.15:74124460 - 74137359 on Build GRCh38 | Deletion | ISLR2 LOC283731 |
esv2760040 | Chr.15:73922773 - 74172575 on Build GRCh38 | Gain+Loss | STOML1 ISLR2 LOXL1 LOC283731 LOC732265 LOXL1-AS1 GOLGA6A PML |
nsv1047564 | Chr.15:74116495 - 74165840 on Build GRCh38 | Gain | ISLR2 LOC283731 |
nsv1121512 | Chr.15:72670085 - 75273877 on Build GRCh38 | Deletion | LOC729739 REC114 PML CYP11A1 ADPGK CYP1A1 ARID3B LOC107984728 SCAMP2 SEMA7A LMAN1L STOML1 ISLR2 RPP25 HCN4 CCDC33 BBS4 NPTN ULK3 LOXL1 C15orf39 ISLR TBC1D21 NPTN-IT1 PPCDC MIR4513 STRA6 LOC283731 MIR6882 GOLGA6A COX5A NEO1 CPLX3 CSK LOC102723750 FAM219B C15orf59 MPI CYP1A2 EDC3 CLK3 LOC105376731 LOC101929333 LOC732265 UBL7-AS1 SCAMP5 MIR6881 C15orf59-AS1 UBL7 CD276 LOXL1-AS1 ADPGK-AS1 GOLGA6C HIGD2B |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |