Assay Details
Target Gene Details
Entrez Gene ID: | 9477 |
Gene Name: | mediator complex subunit 20 |
Gene Aliases: |
PRO0213, TRFP |
Location: |
Chr.6:41905353-41921147 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MED20 | NM_001305455.1 | 3 | 1018 | NP_001292384.1 |
NM_001305456.1 | 5 | 1290 | NP_001292385.1 | |
NM_001305457.1 | 4 | 1097 | NP_001292386.1 | |
NM_004275.4 | 4 | 1173 | NP_004266.2 | |
NR_131160.1 | 3 | 919 | ||
AJ420544.1 | 3 | 976 | ||
AK023092.1 | 4 | 1117 | BAB14399.1 | |
AK097385.1 | 5 | 1290 | ||
AK300573.1 | 3 | 1010 | ||
AK307070.1 | 3 | 911 | ||
AL050196.1 | 2 | 688 | CAB43314.1 | |
BC012618.1 | 4 | 1169 | AAH12618.1 | |
BC019866.1 | 4 | 1150 | ||
BC032552.1 | 4 | 1135 | AAH32552.1 | |
BC040950.1 | 4 | 1109 | AAH40950.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv5278 | Chr.6:41906650 - 41908185 on Build GRCh38 | Insertion | MED20 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |