Assay Details
Target Gene Details
Entrez Gene ID: | 94101 |
Gene Name: | ORMDL sphingolipid biosynthesis regulator 1 |
Gene Aliases: |
- |
Location: |
Chr.2:189766149-189784371 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ORMDL1 | NM_001128150.1 | NP_001121622.1 | ||
NM_016467.4 | NP_057551.1 | |||
DB122266.1 |
Target Gene Details
Entrez Gene ID: | 5378 |
Gene Name: | PMS1 homolog 1, mismatch repair system component |
Gene Aliases: |
HNPCC3, MLH2, PMSL1, hPMS1 |
Location: |
Chr.2:189784085-189877629 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PMS1 | NM_000534.4 | 1 | 320 | NP_000525.1 |
NM_001128143.1 | 1 | 320 | NP_001121615.1 | |
NM_001128144.1 | 1 | 320 | NP_001121616.1 | |
NM_001289408.1 | 1 | 320 | NP_001276337.1 | |
NM_001289409.1 | 1 | 320 | NP_001276338.1 | |
NM_001321044.1 | 1 | 320 | NP_001307973.1 | |
NM_001321045.1 | 1 | 320 | NP_001307974.1 | |
NM_001321046.1 | 1 | 320 | NP_001307975.1 | |
NM_001321047.1 | 1 | 320 | NP_001307976.1 | |
NM_001321048.1 | 1 | 320 | NP_001307977.1 | |
NM_001321049.1 | 1 | 320 | NP_001307978.1 | |
NM_001321051.1 | 1 | 320 | NP_001307980.1 | |
NR_110332.1 | 1 | 320 | ||
CV570894.1 | 1 | 275 | ||
DC347859.1 | 1 | 320 | ||
DC360786.1 |
More Information
Set Membership: |
Intragenic Exonic Non-exonic non-DGV Variation |