Assay Details
Target Gene Details
Entrez Gene ID: | 90632 |
Gene Name: | long intergenic non-protein coding RNA 473 |
Gene Aliases: |
C6orf176, bA142J11.1 |
Location: |
Chr.6:165924048-165988039 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LINC00473 | NR_026860.1 | 1 | 321 | |
NR_026861.1 | 1 | 321 | ||
AK289375.1 | 1 | 205 | ||
BX647106.1 | 1 | 330 | ||
DA416538.1 | 1 | 279 |
Target Gene Details
Entrez Gene ID: | 441177 |
Gene Name: | long intergenic non-protein coding RNA 602 |
Gene Aliases: |
- |
Location: |
Chr.6:165987551-165989615 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LINC00602 | NR_027284.1 | 1 | 169 | |
AK094116.1 | 1 | 169 | BAC04291.1 |
Target Gene Details
Entrez Gene ID: | 10846 |
Gene Name: | phosphodiesterase 10A |
Gene Aliases: |
ADSD2, HSPDE10A, IOLOD, LINC00473, PDE10A19 |
Location: |
Chr.6:165327287-165988121 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PDE10A | XM_011535387.2 | 1 | 403 | XP_011533689.2 |
XM_017010194.1 | 1 | 403 | XP_016865683.1 | |
XM_017010197.1 | 1 | 403 | XP_016865686.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |