Assay Details
Target Gene Details
Entrez Gene ID: | 26097 |
Gene Name: | chromatin target of PRMT1 |
Gene Aliases: |
C1orf77, FL-SRAG, FOP, SRAG, SRAG-3, SRAG-5, pp7704 |
Location: |
Chr.1:153633982-153646306 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CHTOP | NM_001206612.1 | 3 | 525 | NP_001193541.1 |
NM_001244664.1 | 3 | 525 | NP_001231593.1 | |
NM_001317077.1 | 3 | 525 | NP_001304006.1 | |
NM_015607.3 | 3 | 525 | NP_056422.2 | |
XM_017000993.1 | 3 | 484 | XP_016856482.1 | |
AF261137.1 | 3 | 458 | AAG44673.1 | |
AF318362.1 | 3 | 433 | AAL55869.1 | |
AK095230.1 | 2 | 2073 | ||
AK308713.1 | 3 | 432 | ||
AK309844.1 | 3 | 427 | ||
AL512704.1 | 1 | 292 | CAC21650.2 | |
BC059949.1 | AAH59949.1 | |||
BC108721.1 | 2 | 160 | AAI08722.1 | |
BC120961.1 | 3 | 252 | ||
BC120962.1 | 3 | 252 | ||
BP375397.1 | 3 | 432 | ||
DC387712.1 | 3 | 526 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |