Assay Details
Target Gene Details
Entrez Gene ID: | 10574 |
Gene Name: | chaperonin containing TCP1 subunit 7 |
Gene Aliases: |
CCTETA, CCTH, NIP7-1, TCP1ETA |
Location: |
Chr.2:73233431-73253022 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CCT7 | NM_001009570.2 | NP_001009570.1 | ||
NM_001166284.1 | NP_001159756.1 | |||
NM_001166285.1 | NP_001159757.1 | |||
NM_006429.3 | NP_006420.1 | |||
NR_029402.1 | ||||
NR_029403.1 | ||||
XM_011532478.2 | XP_011530780.1 | |||
XM_011532479.1 | XP_011530781.1 | |||
AF026292.1 | AAC96011.1 | |||
AK098549.1 | ||||
AK222478.1 | BAD96198.1 | |||
AK291961.1 | ||||
AK293175.1 | ||||
AK293597.1 | ||||
AK297846.1 | ||||
AK298153.1 | ||||
AK302078.1 | ||||
BC019296.1 | AAH19296.1 | |||
BC088351.1 | AAH88351.1 | |||
CR456719.1 | CAG33000.1 | |||
CR536511.1 | CAG38749.1 | |||
DC362157.1 | ||||
DC395264.1 | ||||
U83843.1 | AAB41437.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv2776 | Chr.2:73204726 - 73255176 on Build GRCh38 | Deletion | NOTO SMYD5 FBXO41 CCT7 PRADC1 |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |