Assay Details
Target Gene Details
Entrez Gene ID: | 84167 |
Gene Name: | chromosome 19 open reading frame 44 |
Gene Aliases: |
- |
Location: |
Chr.19:16496311-16521369 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
C19orf44 | NM_001288834.1 | 8 | 2192 | NP_001275763.1 |
NM_032207.3 | NP_115583.1 | |||
XM_006722920.3 | XP_006722983.1 | |||
XM_011528354.2 | 9 | 2288 | XP_011526656.1 | |
AK025395.1 | BAB15125.1 | |||
AK026145.1 | 5 | 948 | ||
AK225118.1 | 8 | 2109 | ||
AK297781.1 | ||||
BC027869.1 | AAH27869.1 |
Target Gene Details
Entrez Gene ID: | 10523 |
Gene Name: | calcium homeostasis endoplasmic reticulum protein |
Gene Aliases: |
DAN16, SCAF6, SRA1 |
Location: |
Chr.19:16517889-16542452 on Build GRCh38 |
Assay Gene Location: | Within Exon 17 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CHERP | NM_006387.5 | 17 | 3865 | NP_006378.3 |
AA782418.1 | 1 | 241 | ||
AB209730.1 | 8 | 2242 | BAD92967.1 | |
AF536542.1 | 17 | 3832 | AAN77183.1 | |
BC021294.2 | 17 | 3850 | AAH21294.1 | |
U94836.2 | 16 | 3767 | AAB53327.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |