Assay Details
Target Gene Details
Entrez Gene ID: | 90226 |
Gene Name: | urocortin 2 |
Gene Aliases: |
SRP, UCN-II, UCNI, UR, URP |
Location: |
Chr.3:48561718-48563768 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
UCN2 | NM_033199.3 | 2 | 464 | NP_149976.1 |
AF320560.1 | AAK52672.1 | |||
BC002647.2 | 2 | 464 | AAH02647.2 | |
BC022096.1 | 2 | 464 | AAH22096.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv954484 | Chr.3:48533268 - 48609567 on Build GRCh38 | Deletion | UCN2 UQCRC1 MIR711 COL7A1 PFKFB4 MIR6823 SNORA94 |
nsv1004383 | Chr.3:48506361 - 48612339 on Build GRCh38 | Loss | UCN2 UQCRC1 MIR711 COL7A1 PFKFB4 MIR6823 SNORA94 |
nsv3811 | Chr.3:48561747 - 48606376 on Build GRCh38 | Deletion | UCN2 UQCRC1 MIR711 COL7A1 PFKFB4 SNORA94 |
nsv834685 | Chr.3:48527576 - 48708341 on Build GRCh38 | Loss | UCN2 UQCRC1 NCKIPSD CELSR3 SLC26A6 IP6K2 PFKFB4 MIR6823 MIR6824 TMEM89 MIR4793 CELSR3-AS1 MIR711 COL7A1 SNORA94 |
nsv523981 | Chr.3:48478880 - 48683607 on Build GRCh38 | Loss | UCN2 UQCRC1 NCKIPSD CELSR3 SLC26A6 PFKFB4 MIR6823 MIR6824 TMEM89 MIR4793 CELSR3-AS1 SHISA5 MIR711 COL7A1 SNORA94 |
nsv1152618 | Chr.3:48558266 - 48563067 on Build GRCh38 | Duplication | UCN2 PFKFB4 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |