Assay Details
Target Gene Details
Entrez Gene ID: | 8786 |
Gene Name: | regulator of G-protein signaling 11 |
Gene Aliases: |
RS11 |
Location: |
Chr.16:268301-275943 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
RGS11 | NM_001286485.1 | 6 | 462 | NP_001273414.1 |
NM_001286486.1 | 6 | 399 | NP_001273415.1 | |
NM_003834.2 | 7 | 506 | NP_003825.1 | |
NM_183337.2 | 7 | 495 | NP_899180.1 | |
XM_011522719.2 | 8 | 589 | XP_011521021.1 | |
XM_011522720.2 | 8 | 529 | XP_011521022.1 | |
XM_011522721.1 | 8 | 589 | XP_011521023.1 | |
XM_011522722.1 | 8 | 589 | XP_011521024.1 | |
XM_011522723.1 | 7 | 493 | XP_011521025.1 | |
AB016929.1 | 6 | 430 | BAA74751.1 | |
AB209582.1 | 2 | 948 | BAD92819.1 | |
AF035153.1 | 7 | 466 | AAC69175.1 | |
AF035154.1 | 7 | 477 | AAC69176.1 | |
AK091701.1 | 6 | 396 | ||
AK293564.1 | 7 | 489 | ||
AK294448.1 | 7 | 489 | ||
AK308972.1 | 6 | 716 | ||
BC040504.1 | 6 | 1243 | AAH40504.1 | |
BC141803.1 | 7 | 470 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2422427 | Chr.16:10001 - 2723164 on Build GRCh38 | Duplication | PDIA2 RHOT2 SNORA64 NME3 MPG MIR3677 GNG13 ARHGDIG LOC107984876 MIR3177 LOC107987233 RAB11FIP3 ERVK13-1 PKD1 MIR4717 TPSB2 BRICD5 MIR1225 MCRIP2 RNF151 TMEM8A MIR6511B1 CEMP1 DDX11L10 LOC106660606 SPSB3 LUC7L SSTR5-AS1 CLCN7 KCTD5 AMDHD2 ZNF598 BAIAP3 ABCA17P TMEM204 TELO2 DECR2 CHTF18 LOC105371047 IFT140 NARFL TSR3 MIR940 LOC105371184 GNPTG CRAMP1 JMJD8 MIR3176 LOC105371038 NPW SOX8 LMF1 ABCA3 GFER SSTR5 RNPS1 PRR25 MRPS34 AXIN1 UBE2I LOC105371049 NME4 HBZ CCNF LINC00254 MIR4516 CCDC78 TPSAB1 HS3ST6 PTX4 NUBP2 C1QTNF8 STUB1 LINC00235 LOC652276 LOC105371046 FBXL16 METTL26 TBC1D24 MAPK8IP3 CASKIN1 MIR3178 FAM173A HBQ1 NHLRC4 MIR6859-4 SNRNP25 NPRL3 HBA1 EME2 RHBDF1 SNHG19 HN1L ECI1 RAB26 HBA2 LOC105371045 TPSG1 IGFALS HBM FAM234A SYNGR3 WDR90 TPSD1 SNORA10 HAGHL SNORD60 MSLN LOC729652 PRSS29P NDUFB10 NOXO1 C16orf59 ATP6V0C HAGH POLR3K MIR5587 WASIR2 METRN TRAF7 CACNA1H MSRB1 SNHG9 LMF1-AS1 WFIKKN1 SNORA78 RHBDL1 FAHD1 TBL3 PRSS27 MLST8 MEIOB SLC9A3R2 RPUSD1 E4F1 RPL23AP5 RPS2 MRPL28 CCDC154 LOC100134368 NTHL1 DNASE1L2 PRR35 LOC100287175 RGS11 CAPN15 PDPK1 PGP WDR24 RAB40C TSC2 RPL3L MIR3180-5 UNKL PIGQ MIR6767 C16orf91 FLJ42627 NTN3 MIR6768 MIR662 LOC101929280 |
esv3570031 | Chr.16:243389 - 278107 on Build GRCh38 | Gain | RGS11 FAM234A |
nsv952896 | Chr.16:273501 - 321500 on Build GRCh38 | Deletion | PDIA2 RGS11 AXIN1 ARHGDIG |
esv3637563 | Chr.16:263248 - 505590 on Build GRCh38 | Gain | PDIA2 NME4 TMEM8A RAB11FIP3 RGS11 RPL23AP5 MRPL28 FAM234A DECR2 AXIN1 ARHGDIG LOC100134368 |
nsv827477 | Chr.16:262247 - 297639 on Build GRCh38 | Gain | PDIA2 RGS11 FAM234A AXIN1 ARHGDIG |
nsv570947 | Chr.16:272935 - 297326 on Build GRCh38 | Loss | PDIA2 RGS11 AXIN1 ARHGDIG |
dgv32n68 | Chr.16:245507 - 476226 on Build GRCh38 | Loss | PDIA2 NME4 TMEM8A RAB11FIP3 RGS11 RPL23AP5 MRPL28 FAM234A DECR2 AXIN1 ARHGDIG LOC100134368 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |