Assay Details
Target Gene Details
Entrez Gene ID: | 6284 |
Gene Name: | S100 calcium binding protein A13 |
Gene Aliases: |
- |
Location: |
Chr.1:153618799-153634092 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
S100A13 | NM_001024210.1 | NP_001019381.1 | ||
NM_001024211.1 | NP_001019382.1 | |||
NM_001024212.1 | NP_001019383.1 | |||
NM_001024213.1 | NP_001019384.1 | |||
NM_005979.2 | NP_005970.1 | |||
XM_005245434.3 | XP_005245491.1 | |||
XM_011509862.2 | XP_011508164.1 | |||
XM_011509863.2 | XP_011508165.1 | |||
XM_011509864.1 | XP_011508166.1 | |||
XM_017002034.1 | XP_016857523.1 | |||
XM_017002035.1 | XP_016857524.1 | |||
XM_017002036.1 | XP_016857525.1 | |||
AK097132.1 | ||||
AY987392.1 | AAX89402.1 | |||
BC000632.2 | AAH00632.1 | |||
BC068064.1 | AAH68064.1 | |||
BC070291.1 | AAH70291.1 | |||
BM762088.1 | ||||
BT006724.1 | AAP35370.1 | |||
BX472819.1 | ||||
CR542149.1 | CAG46946.1 | |||
X99920.1 | CAA68188.1 |
More Information
Set Membership: |
Intragenic Non-exonic non-DGV Variation |