Assay Details
Target Gene Details
Entrez Gene ID: | 22858 |
Gene Name: | intestinal cell (MAK-like) kinase |
Gene Aliases: |
ECO, LCK2, MRK |
Location: |
Chr.6:53001299-53061864 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ICK | NM_014920.3 | 14 | 5754 | NP_055735.1 |
NM_016513.4 | 15 | 5865 | NP_057597.2 | |
XM_011514419.1 | 14 | 5775 | XP_011512721.1 | |
XM_011514420.2 | 15 | 5917 | XP_011512722.1 | |
XM_011514421.1 | 15 | 5856 | XP_011512723.1 | |
XM_017010485.1 | 15 | 6149 | XP_016865974.1 | |
XM_017010486.1 | 16 | 6028 | XP_016865975.1 | |
XM_017010487.1 | 14 | 5694 | XP_016865976.1 | |
XM_017010488.1 | 15 | 6555 | XP_016865977.1 | |
XM_017010489.1 | 16 | 6070 | XP_016865978.1 | |
XM_017010490.1 | 15 | 5940 | XP_016865979.1 | |
XM_017010491.1 | 15 | 5901 | XP_016865980.1 | |
XM_017010492.1 | 14 | 5945 | XP_016865981.1 | |
AB023153.1 | 14 | 5641 | ||
AJ420557.1 | 1 | 1642 | ||
BC152464.1 | 14 | 5641 | ||
BX647493.1 | 2 | 3997 |
More Information
Additional Information:
For this assay, SNP(s) [rs151300001] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
Intragenic Exonic non-DGV Variation |