Assay Details
Target Gene Details
Entrez Gene ID: | 7917 |
Gene Name: | BCL2 associated athanogene 6 |
Gene Aliases: |
BAG-6, BAT3, D6S52E, G3 |
Location: |
Chr.6:31639028-31660900 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
BAG6 | XM_017011279.1 | 1 | 130 | XP_016866768.1 |
Target Gene Details
Entrez Gene ID: | 57827 |
Gene Name: | chromosome 6 open reading frame 47 |
Gene Aliases: |
D6S53E, G4, NG34 |
Location: |
Chr.6:31658298-31660772 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
C6orf47 | NM_021184.3 | NP_067007.3 | ||
BC012950.1 | AAH12950.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv462871 | Chr.6:31607499 - 31664357 on Build GRCh38 | Loss | GPANK1 MIR6832 APOM C6orf47 PRRC2A AIF1 BAG6 SNORA38 |
nsv1126749 | Chr.6:31457323 - 32594423 on Build GRCh38 | Deletion | ABHD16A PPT2-EGFL8 PPT2 SNORD52 GPANK1 APOM HLA-DRA LY6G6D EHMT2 LOC100507547 SNORD48 NEU1 CYP21A1P LOC101929163 SNORD84 DDAH2 HLA-DRB5 CSNK2B C6orf48 MIR6833 HCP5 SKIV2L MIR4646 ATP6V1G2-DDX39B DXO LTA MCCD1 VWA7 LY6G6F VARS HSPA1A LOC102725068 C6orf25 TNXB CLIC1 FKBPL STK19 LY6G5C GPSM3 LOC100287329 EGFL8 SNORD117 MIR6721 CYP21A2 PRRC2A AGPAT1 LSM2 LST1 BAG6 MICB SLC44A4 CFB MIR6832 NELFE BTNL2 C4B C4A SAPCD1-AS1 LTB HSPA1L LY6G6E AIF1 HLA-DRB6 DDX39B-AS1 HLA-DRB1 ZBTB12 LY6G6C PBX2 SAPCD1 MSH5-SAPCD1 TNXA C2-AS1 TNF RNF5 C2 NOTCH4 HSPA1B C6orf47 LY6G5B HCG23 AGER C6orf10 HCG26 NFKBIL1 NCR3 MSH5 DDX39B ATP6V1G2 PRRT1 SNORA38 ATF6B MIR1236 |
nsv1073969 | Chr.6:31456422 - 33480523 on Build GRCh38 | Deletion | SNORD52 APOM HLA-DRA RING1 MIR3135B EHMT2 LOC100507547 NEU1 HLA-DRB5 HLA-DOA CSNK2B HLA-DOB SKIV2L ATP6V1G2-DDX39B HLA-DQB1-AS1 HLA-DQB2 LY6G6F VARS BRD2 LOC102725068 LOC100294145 PFDN6 HLA-DPB1 CLIC1 FKBPL STK19 HSD17B8 LY6G5C LOC100287329 EGFL8 SNORD117 MIR6721 CYP21A2 PRRC2A AGPAT1 LSM2 KIFC1 MIR6873 CFB MIR6832 BTNL2 SAPCD1-AS1 HSPA1L WDR46 HLA-DMA HLA-DRB1 PPP1R2P1 HCG24 MSH5-SAPCD1 TNXA HCG23 AGER C6orf10 PSMB8 HLA-DPA1 DDX39B PSMB9 ATF6B MIR1236 ABHD16A PPT2-EGFL8 PPT2 GPANK1 COL11A2 LY6G6D SNORD48 HLA-DQB1 MIR5004 CYP21A1P LOC101929163 RXRB SNORD84 DDAH2 C6orf48 MIR6833 HCP5 DAXX HLA-DMB MIR219A1 MIR4646 DXO RPS18 LTA MCCD1 VWA7 HSPA1A TAP1 C6orf25 TNXB GPSM3 SYNGAP1 PSMB8-AS1 LST1 BAG6 MICB ZBTB9 SLC44A4 CUTA NELFE RGL2 C4B C4A SLC39A7 LTB LY6G6E AIF1 TAP2 HLA-DRB6 DDX39B-AS1 B3GALT4 ZBTB12 MIR6834 HLA-DPB2 LY6G6C PBX2 SAPCD1 TAPBP PHF1 C2-AS1 TNF RNF5 C2 NOTCH4 HSPA1B HLA-DQA2 ZBTB22 C6orf47 LY6G5B HCG26 NFKBIL1 HLA-DQA1 NCR3 MSH5 ATP6V1G2 HCG25 PRRT1 SNORA38 VPS52 |
dgv10403n54 | Chr.6:31312324 - 32049763 on Build GRCh38 | Loss | ABHD16A SNORD52 GPANK1 APOM LY6G6D EHMT2 SNORD48 MIR6891 NEU1 CYP21A1P SNORD84 DDAH2 CSNK2B C6orf48 HCP5 SKIV2L MIR4646 ATP6V1G2-DDX39B DXO LTA MCCD1 VWA7 LY6G6F VARS HSPA1A LOC102725068 C6orf25 TNXB CLIC1 STK19 LY6G5C LOC100287329 SNORD117 CYP21A2 PRRC2A LSM2 LST1 BAG6 MICB MICA SLC44A4 CFB MIR6832 NELFE C4B C4A SAPCD1-AS1 LTB HSPA1L LY6G6E AIF1 DDX39B-AS1 ZBTB12 LY6G6C SAPCD1 HLA-B MSH5-SAPCD1 TNXA C2-AS1 TNF C2 HSPA1B C6orf47 LY6G5B HCG26 NFKBIL1 NCR3 MSH5 DDX39B ATP6V1G2 SNORA38 MIR1236 |
nsv1112900 | Chr.6:31455323 - 33480423 on Build GRCh38 | Deletion | SNORD52 APOM HLA-DRA RING1 MIR3135B EHMT2 LOC100507547 NEU1 HLA-DRB5 HLA-DOA CSNK2B HLA-DOB SKIV2L ATP6V1G2-DDX39B HLA-DQB1-AS1 HLA-DQB2 LY6G6F VARS BRD2 LOC102725068 LOC100294145 PFDN6 HLA-DPB1 CLIC1 FKBPL STK19 HSD17B8 LY6G5C LOC100287329 EGFL8 SNORD117 MIR6721 CYP21A2 PRRC2A AGPAT1 LSM2 KIFC1 MIR6873 CFB MIR6832 BTNL2 SAPCD1-AS1 HSPA1L WDR46 HLA-DMA HLA-DRB1 PPP1R2P1 HCG24 MSH5-SAPCD1 TNXA HCG23 AGER C6orf10 PSMB8 HLA-DPA1 DDX39B PSMB9 ATF6B MIR1236 ABHD16A PPT2-EGFL8 PPT2 GPANK1 COL11A2 LY6G6D SNORD48 HLA-DQB1 MIR5004 CYP21A1P LOC101929163 RXRB SNORD84 DDAH2 C6orf48 MIR6833 HCP5 DAXX HLA-DMB MIR219A1 MIR4646 DXO RPS18 LTA MCCD1 VWA7 HSPA1A TAP1 C6orf25 TNXB GPSM3 SYNGAP1 PSMB8-AS1 LST1 BAG6 MICB ZBTB9 SLC44A4 CUTA NELFE RGL2 C4B C4A SLC39A7 LTB LY6G6E AIF1 TAP2 HLA-DRB6 DDX39B-AS1 B3GALT4 ZBTB12 MIR6834 HLA-DPB2 LY6G6C PBX2 SAPCD1 TAPBP PHF1 C2-AS1 TNF RNF5 C2 NOTCH4 HSPA1B HLA-DQA2 ZBTB22 C6orf47 LY6G5B HCG26 NFKBIL1 HLA-DQA1 NCR3 MSH5 ATP6V1G2 HCG25 PRRT1 SNORA38 VPS52 |
dgv10404n54 | Chr.6:31312550 - 32516591 on Build GRCh38 | Loss | ABHD16A PPT2-EGFL8 PPT2 SNORD52 GPANK1 APOM HLA-DRA LY6G6D EHMT2 LOC100507547 SNORD48 MIR6891 NEU1 CYP21A1P LOC101929163 SNORD84 DDAH2 CSNK2B C6orf48 MIR6833 HCP5 SKIV2L MIR4646 ATP6V1G2-DDX39B DXO LTA MCCD1 VWA7 LY6G6F VARS HSPA1A LOC102725068 C6orf25 TNXB CLIC1 FKBPL STK19 LY6G5C GPSM3 LOC100287329 EGFL8 SNORD117 MIR6721 CYP21A2 PRRC2A AGPAT1 LSM2 LST1 BAG6 MICB MICA SLC44A4 CFB MIR6832 NELFE BTNL2 C4B C4A SAPCD1-AS1 LTB HSPA1L LY6G6E AIF1 DDX39B-AS1 ZBTB12 LY6G6C PBX2 SAPCD1 HLA-B MSH5-SAPCD1 TNXA C2-AS1 TNF RNF5 C2 NOTCH4 HSPA1B C6orf47 LY6G5B HCG23 AGER C6orf10 HCG26 NFKBIL1 NCR3 MSH5 DDX39B ATP6V1G2 PRRT1 SNORA38 ATF6B MIR1236 |
nsv1161370 | Chr.6:31600692 - 31714252 on Build GRCh38 | Deletion | ABHD16A GPANK1 MIR6832 MIR4646 APOM C6orf47 LY6G6F LY6G6E LY6G5B AIF1 CSNK2B LY6G5C PRRC2A BAG6 SNORA38 |
nsv830627 | Chr.6:31528437 - 31692173 on Build GRCh38 | Loss | ABHD16A GPANK1 MIR6832 APOM LTB AIF1 DDX39B-AS1 SNORD84 CSNK2B TNF ATP6V1G2-DDX39B C6orf47 LTA MCCD1 LY6G5B NFKBIL1 NCR3 LY6G5C LOC100287329 SNORD117 DDX39B ATP6V1G2 PRRC2A LST1 BAG6 SNORA38 |
More Information
Set Membership: |
Intragenic Exonic Non-exonic DGV Variation |